Differentiated NG-108 cells loaded with FluoVolt™ Membrane Potential Dye
Cells were imaged with 10 millisecond illumination pulses and images acquired with 2X binning. The three selected traces (B) show fluorogenic responses from the dye as selected cells (A) spontaneously depolarize and repolarize in culture.
Cells were imaged with 10 millisecond illumination pulses and images acquired with 2X binning. The three selected traces (B) show fluorogenic responses from the dye as selected cells (A) spontaneously depolarize and repolarize in culture.
Related Products
Related Images