Fluorescence immunostaining of a fixed co-culture of murine neurons and dendritic cells.
Fluorescence immunostaining of a fixed co-culture of murine neurons and dendritic cells. Neurons were labeled with an antibody directed against neurofilament triplet H (NFH) protein and visualized using red-fluorescent tetramethylrhodamine goat anti–mouse IgG (CAt. no. T2762). Dendritic cells were labeled with a biotin conjugate of a primary antibody directed against the integrin CD11c and visualized using green-fluorescent Alexa Fluor® 488 streptavidin (Cat. no. S11223, S32354). Nuclei were stained with blue-fluorescent TO-PRO®-3 (Cat. no. T3605). Image submitted by Gauthier Dorban, Institute of Human Histology, University of Liège, Belgium.
Fluorescence immunostaining of a fixed co-culture of murine neurons and dendritic cells. Neurons were labeled with an antibody directed against neurofilament triplet H (NFH) protein and visualized using  red-fluorescent tetramethylrhodamine goat anti–mouse IgG (CAt. no. T2762). Dendritic cells were labeled with a biotin conjugate of a primary antibody directed against the integrin CD11c and visualized using green-fluorescent Alexa Fluor® 488 streptavidin (Cat. no. S11223, S32354). Nuclei were stained with blue-fluorescent TO-PRO®-3 (Cat. no. T3605). Image submitted by Gauthier Dorban, Institute of Human Histology, University of Liège, Belgium.
Related Products
Related Images