GLA Antibody (PA5-13687) in FACS
Flow cytometry analysis of HepG2 cells using a GLA polyclonal antibody (Product ## PA5-13687) (right) compared to a negative control cell (left) at a dilution of 1:10-50, followed by a FITC-conjugated goat anti-rabbit antibody
Flow cytometry analysis of HepG2 cells using a GLA polyclonal antibody (Product ## PA5-13687) (right) compared to a negative control cell (left) at a dilution of 1:10-50, followed by a FITC-conjugated goat anti-rabbit antibody
Related Products
Related Images