Heratherm Incubators
Your biological samples are often irreplaceable. That makes your microbiological incubator critical to your success. Your work is challenging enough. Surround your life with some certainty with Thermo Scientific Heratherm microbiological incubators—available in four models, providing three incubator airflow technologies in a choice of six sizes.   
Download brochure
Select from our basic compact option or line of advanced incubators.
Our portfolio of Heratherm incubators are designed to maintain sample integrity and temperature consistency - for microbiological incubation, growth of yeast or other microorganisms, storage and testing. These microbiological incubators are available in various configurations and six different sizes to suit every lab's needs.
  Compact
Incubator
General  Protocol
Incubators
Advanced Protocol 
Incubators
Advanced Protocol
Security Incubators
    Table Top Large Capacity   Table Top Large Capacity
Temperature range +18° to +40℃ ambient
+5℃ to +75℃
ambient
+5℃ to +105℃
ambient
+5℃ to +105℃
Convection technology Peltier with Mechanical  Gravity Dual Dual Mechanical
Automatic over-temperature alarm  
Automatic under-temperature alarm - - -
Timer - on / off weekly/daily/real time weekly/daily/real time
Certified decontamination
cycle
- - -
Door alarm - - -
Lockable door - - -
Connection for optional sample temperature sensor - - -
Featured Video
Heratherm microbiological incubators
Watch this video to learn how our Heratherm microbiological incubators can meet your application needs.