Search
Thermo Scientific HyperSep microscale solid phase extraction (MSPE) Tips and SpinTips were designed to solve the difficulties of performing SPE sample preparation on small-volume samples. Using a patented design, these Tips and SpinTips are available with a variety of SPE resins to satisfy a wide range of low-sample-volume applications. Use these tips for chromatography, mass spectrometry, desalting, protein purification, and electrophoresis.
SPE media in Thermo Scientific HyperSep Microscale Solid Phase Extraction Tips is directly attached to the inner surface of micropipette tip without using polymers or glue. This design avoids potential problems with contamination or permeability, and maximizes the surface area of the chromatographic media in contact with the sample.
| Stationary Phase | 1-10μL Capacity | 10-200μL Capacity | Quantity |
|---|---|---|---|
| BioBasic C18 | 60109-201 | 60109-209 | 96 Pack |
| BioBasic C8 | 60109-202 | 60109-210 | 96 Pack |
| BioBasic C4 | 60109-203 | 60109-211 | 96 Pack |
| Hypercarb | 60109-204 | 60109-212 | 96 Pack |
| Hypercarb & C18 Mixed Mode | 60109-205 | 60109-213 | 96 Pack |
| HILIC | 60109-206 | 60109-214 | 96 Pack |
| Trypsin | 60109-207 | 60109-215 | 96 Pack |
| Titanium Dioxide | 60109-208 | 60109-216 | 96 Pack |
| Zirconium Dioxide | 60109-217 | 60109-218 | 96 Pack |
Thermo Scientific HyperSep SpinTips are another ideal option for perform SPE sample preparation on low-volume samples. SPE materials reside in the pipette tip, which has a fine slit at the bottom (slit width 1-2 µm) to permit liquid to pass through while the chromatographic material (20-30 µm) remains in the tip. This also eliminates the need for a filter and thus reduces dead volume.
| Stationary Phase | 1-10μL Capacity Tips | 10-200μL Capacity Tips | Quantity |
|---|---|---|---|
| C18 | 60109-401 | 60109-412 | 96 Pack |
| C8 | 60109-402 | 60109-413 | 96 Pack |
| C4 | 60109-403 | 60109-414 | 96 Pack |
| Hypercarb | 60109-404 | 60109-415 | 96 Pack |
| Hypercarb & C18 Mixed Mode | 60109-405 | 60109-416 | 96 Pack |
| HILIC | 60109-406 | 60109-417 | 96 Pack |
| Trypsin | 60109-407 | 60109-418 | 96 Pack |
| POROS WAX Weak Anion Exchanger | 60109-408 | 60109-419 | 96 Pack |
| POROS SAX Strong Anion Exchanger | 60109-409 | 60109-420 | 96 Pack |
| POROS SCX Strong Cation Exchanger | 60109-410 | 60109-421 | 96 Pack |
| Titanium Dioxide | 60109-411 | 60109-422 | 96 Pack |
| Zirconium Dioxide | 60109-424 | 60109-425 | 96 Pack |
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.