Which EVOS imaging system is right for you?
All EVOS imaging systems share common features: high-quality LED light sources, cameras, and optics for high-resolution imaging; efficient, simple-to-use design for minimal handling and faster results; powerful, intuitive software for enhanced productivity and reproducibility.
We offer three traditional digital microscopes, and one spectral imaging system designed for spatial proteomics. The EVOS M3000 Imaging System is a manual microscope designed for basic cell culture applications. The EVOS M5000 Imaging System is a semi-automated microscope designed to excel in both cell culture and imaging, providing a flexible solution for diverse research needs. The EVOS M7000 Imaging System is a fully automated microscope designed to simplify the most demanding tissue and cell imaging applications. The EVOS S1000 Spatial Imaging System performs 9-plex tissue imaging with automatic spectral unmixing in a single round without bleaching or cycling.
Choose the right system based on your research needs and applications.
EVOS imaging system selection guide
EVOS M3000 system
EVOS M5000 system
EVOS M7000 system
EVOS S1000 system
Imaging mode
Fluorescence, phase-contrast, brightfield, and color imaging
Sample
Cell  Cell & tissue
Cell & tissue
Tissue
Maximum number of fluorescent colors*
2 4 4 9
Stage operation
Manual Manual Motorized Motorized
Automated image acquisition and autofocus
No No Yes Yes
*In a single round of imaging
Hardware and optics
EVOS M3000 EVOS M5000 EVOS M7000 EVOS S1000
Simple Installation
Installation and training
User User or Service Team
Service Team
Service Team
Motorized encoded X/Y scanning stage
--- ---
Manual mechanical stage (optional)
--- ---
Objective turret positions
4 5 5 5
Phase contrast
Objective type
EVOS, Olympus EVOS, Olympus EVOS, Olympus Zeiss
Objective range
1.25-60x 1.25-100x 1.25- 100X 2.5-40x
Fluorescence channels
2 4 4 9
Light source
Fluorescence LED light cubes Fluorescence LED light cubes Fluorescence LED light cubes Custom LED engine
Monochrome or color camera option (built-in, not optional)
Color camera Mono with LED-based RGB illumination scheme Both Monochrome sCMOS camera
Epifluorescence images
Transmitted-light images
Color images
Benchtop system
Suitable for use in tissue culture hood
--- ---
Darkroom needed
No No No No
Onstage incubator for time-lapse imaging
--- Optional Optional ---
Autofocus
--- Multichannel Multichannel Custom IR laser focus and software autofocus
Z-stack capability
--- ---
Automated multiwell plate screening
--- --- ---
Cloud connectivity
USB Ports
External monitor support
Software attributes
EVOS M3000 EVOS M5000 EVOS M7000 EVOS S1000
Celleste analysis software
--- Optional Optional ---
Automatic cell counting
--- ---
Cell confluency 
---
Transfection efficiency
--- ---
Stage tracking
---
Intuitive onboard software
Networking capability
Spectral unmixing 
--- --- ---
Reagents for EVOS
For Research Use Only. Not for use in diagnostic procedures.