Search
A culture of Saccharomyces cerevisiae is incubated in medium containing the FUN® 1 viability indicator (Cat. No. F7030) and the counterstain Calcofluor White M2R, both of which are provided in our LIVE/DEAD® Yeast Viability Kit (Cat. No. L7009). Metabolically active yeast process the FUN® 1 dye, forming numerous red-fluorescent cylindrical structures within their vacuoles. Calcofluor stains the cell walls fluorescent blue, regardless of the yeast's metabolic state. The yeast were photographed in a single exposure through an Omega Optical triple bandpass filter set.
Live cell imaging with CellLight™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›
Staining with MDY-64. Go ›
Staining with Arg-CMAC. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›