Search
The smart design of the Ion Torrent Genexus Dx Integrated Sequencer is designed to reduce errors while increasing your lab’s productivity.
With a single touchpoint and as little as ten minutes of hands-on time, the Ion Torrent Genexus Dx Integrated Sequencer starts with purified nucleic acid and automates all remaining steps of the targeted next-generation sequencing (NGS) workflow, including cDNA synthesis and library preparation, template preparation, sequencing, data analysis, and variant reporting. The result is an efficient workflow that is easily implemented in a variety of clinical lab settings.
The user can select their desired sample(s) and assay(s) and use the intuitive setup wizard to create a run plan for the sequencer.
The onboard vision system verifies consumable installation in real-time using barcode and RFID detection, enabling proper setup in minutes.
The user can view a report for each sample, sample-level quality control metrics, and a list of reagents that were used.
The Genexus Dx Integrated Sequencer features dual-mode software enabling use of the instrument for both diagnostic (IVD) and research (assay development) applications. You even have the option to multiplex up to 32 library reactions per run to increase sample throughput or to analyze individual samples as they come in to decrease turnaround time.
The Genexus Dx Integrated Sequencer enhances your laboratory's productivity and innovation, whether you are running diagnostic tests or performing clinical research. It is ready to help testing labs in the United States and other countries deliver critical genomic information to patients and their healthcare team.
Intended Use: The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.
Stringent performance characteristics to enable diagnostic use
Developed under design control principles, compliant with US Food & Drug Administration (FDA) Quality System Regulation (QSR) 21 CFR Part 820, and manufactured in facilities registered with FDA, ISO13485-certified, and under Current Good Manufacturing Practices (cGMPs).
![]()
Perform IVD assays and conduct clinical research on the same instrument
Both IVD and RUO assays can be run on the Genexus Dx Integrated sequencer using the dual mode Genexus Dx Software.
Modulate run setup as samples come in with the flexibility of multiplexing up to 32 library reactions per run.
A Genexus Dx Integrated Sequencer run starts with purified nucleic acid and automates all remaining steps of the targeted NGS workflow.
Automated library prep, sequencing, and reporting all happen on one instrument with a set-up-and-go workflow that includes only a single touchpoint and as little as 10 minutes of hands-on time.
For In Vitro Diagnostic Use.
CN: 63522