IL-13 Receptor alpha 2 Antibody (PA5-46976) in WB
Western blot analysis from lysates of human placenta tissue and human testis tissue, loaded at 0.2 mg/mL. A specific band was detected for IL-13 Rα2 at approximately 40 kDa (as indicated) using 10 µg/mL of Goat Anti-human IL-13 Rα2 Antigen Affinity-purified Polyclonal Antibody (Product # PA5-46976).
Western blot analysis from lysates of human placenta tissue and human testis tissue, loaded at 0.2 mg/mL. A specific band was detected for IL-13 Rα2 at approximately 40 kDa (as indicated) using 10 µg/mL of Goat Anti-human IL-13 Rα2 Antigen Affinity-purified Polyclonal Antibody (Product # PA5-46976).
Related Products
Related Images