TMEM184A Antibody (PA564460) in IHC
Immunohistochemical staining of TMEM184A in human tonsil tissue shows mmembane and cytoplasmic positivity in a subset of leukocytes. Samples were probed using a TMEM184A Polyclonal Antibody (Product # PA5-64460).
Immunohistochemical staining of TMEM184A in human tonsil tissue shows mmembane and cytoplasmic positivity in a subset of leukocytes. Samples were probed using a TMEM184A Polyclonal Antibody (Product # PA5-64460).
Related Products
Related Images