B·R·A·H·M·S PCT-Q
Thermo Scientific™ B·R·A·H·M·S™ PCT-Q is an immunochromatographic point-of-care test for the determination of PCT (procalcitonin) in serum and plasma.
B·R·A·H·M·S PCT-Q is CE marked.
Assay Characteristics
The B·R·A·H·M·S PCT-Q assay can be distinguished by these characteristics:
Reliability and reproducibility
Thermo Scientific B·R·A·H·M·S PCT-Q shows a high degree of reliability and reproducibility in the determination of PCT concentrations in serum and plasma. Semi-quantitative concentration ranges obtained with B·R·A·H·M·S PCT-Q correlate closely with the quantitative results obtained with B·R·A·H·M·S PCT LIA.
B·R·A·H·M·S PCT-Q achieves 90-92% of diagnostic sensitivity and 92-98% of diagnostic specificity when compared with the clinical use of B·R·A·H·M·S PCT LIA.
Assay procedure
  • Pipette 200 μL of serum/plasma into the round cavity. Document the time on the reference card.
  • Incubate for 30 minutes at room temperature.
  • After 30 minutes (max. 45 minutes), determine the PCT concentration range of the sample by comparing the color intensity of the test band with the color blocks of the reference card.
PCT
  1. No band or only test band visible: tests which show no control band are not valid and may not be evaluated.
  2. Only control band visible: tests which show only a control band are negatively valid. The PCT concentration is <0.5 μg/L.
  3. Control and test band visible: tests which show both a control band and a test band are positively valid.
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country. Availability of product in each country depends on local regulatory marketing authorization status.