Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 4 | 561 | 590/40 | 551 | 614 | (in buffer) 5 | flow cytometry |
Invitrogen NovaFluor Yellow 610 dye can be used to replace PE/Dazzle™ 594, BD Horizon™ PE-CF594, or PE-Texas Red dye using the 561 nm yellow laser line for excitation. In comparison to these PE tandem dyes, NovaFluor Yellow 610 dye shows much less cross-excitation and lower spectral spillover with the 488 nm blue laser line, allowing detection of an additional marker labeled with, for example, one of the NovaFluor Blue 610 dyes.
The macromolecule-based NovaFluor Yellow 610 dye produces highly stable fluorescence, and stained samples retain their fluorescence intensity and spectral signature when stored at 4°C. Use NovaFluor dyes with CellBlox Plus Blocking Buffer to reduce background and to block non-specific binding of NovaFluor labels, PE and APC tandems observed with macrophages and monocytes.
We offer NovaFluor dyes conjugated to primary antibodies for use in flow cytometry, as well as NovaFluor Antibody Conjugation Kits, NovaFluor CD4 Label Characterization Kits, and custom conjugation services.
Spectral signature of NovaFluor Yellow 610 dye. Normal human peripheral blood cells stained with CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor Yellow 610 were used for analysis. Data were acquired on a 5-laser Cytek Aurora system.
Staining of human peripheral blood cells with NovaFluor Yellow 610 dye conjugated antibody. Normal human peripheral blood cells (PBMCs) were unstained (left) or stained with CD16 Monoclonal Antibody (3G8), NovaFluor Yellow 610 (right). Data were acquired in the YG3 channel on a 5-laser Cytek Aurora system, and singlet cells were used for analysis.
NovaFluor dyes are built using Phiton technology and are compatible with both spectral flow cytometry and traditional flow cytometry. NovaFluor dyes exhibit narrow emission spectra and minimal cross-laser excitation, which helps reduce spectral spillover for better marker resolution. In addition, their unique spectral signatures can provide the opportunity to detect additional markers in flow cytometry panels by opening up previously unusable channels.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.