Search
This product is being discontinued and replaced with the 1500 Series A2 BSC.
Thermo Scientific 1300 Series Class II, Type A2 biological safety cabinets combine safety, energy efficiency and value for today’s laboratory. The cabinet is energy efficient and provides both personal and product protection from biological hazards and contamination. It can be connected to an external exhaust to allow work with volatile toxic chemicals that are sometimes used in the cabinet.
Powering the Thermo Scientific SmartFlow airflow compensation system while delivering better energy efficiency and quieter operation. Night setback mode furthers energy savings and system longevity.
Easily access the interior of the BSC by sliding down the front window.
Choose from work surface designs that best suit your application and work style. Standard equipment includes 2 Duplex receptacles and 6 ports for service valves (3 on each side).
Extends bulb life, saves energy and reduces maintenance.
Clean and safe method of routing vacuum tubing and cables through the side walls, located on left and right sides.
3-, 4-, 5-, or 6-foot sizes.
All applications where personnel, environmental and product protection is required, as well as protection from cross-contamination.
Typical markets: Academia, Clinical, Biotech
Typical applications: cell culture, clinical research, life science research, microbiology, healthcare
NSF/ANSI 49, UL, CE
Cabinet width |
Sash opening choices |
Chamber material choices |
See ordering details |
3 ft |
8 in 10 in |
Stainless steel SmartCoat |
|
4 ft |
8 in 10 in |
Stainless steel SmartCoat
|
Cabinet with accessories package
|
5 ft |
8 in 10 in |
Stainless steel SmartCoat
|
Cabinet with accessories package
|
6 ft |
8 in 10 in |
Stainless steel SmartCoat
|
Cabinet with accessories package
|