Search
The iBind™ Western System is an automated western blot processing platform that requires less primary antibody and delivers sensitive, reproducible western results. All blocking, antibody incubation, and washing steps are hands-free, allowing you to load your solutions and walk away. There is no electricity or battery required. You can also use your existing chemiluminescent, chromogenic, or fluorescent western detection protocols, including HRP-, AP-, Alexa Fluor® dye–, and IRDye® (LI-COR®)–conjugated secondary antibodies.
This cost calculator is designed to help illustrate potential cost savings based on simple and common cost factors. The default values are based on common costs but can be changed as needed to reflect individual customer conditions.
If you have questions or concerns about this tool please email priya.rangaraj@thermofisher.com