Search
Primary XPS region: Ne1s
Overlapping regions: F KLL
Binding energies of common chemical states:
| Chemical state | Binding energy Ne1s |
|---|---|
| Implanted Ne | 861.9 eV |
Experimental information
Interpretation of XPS spectra
N/A
About this element
Symbol: Ne
Date of discovery: 1898
Name origin: Greek neon
Appearance: colorless
Discoverer: Sir William Ramsay, Morris T. Travers
Obtained from: extracted from air
Melting point: -248.6 K
Boiling point: -246.1 K
Density[kg/m3]: unknown
Molar volume: 13.23 × 10-6 m3/mol
Protons/Electrons: 10
Neutrons: 10
Shell structure: 2,8
Electron configuration: [He]2s22p6
Oxidation state: unknown
Crystal structure: cubic
Neon was discovered through the study of liquefied air by British chemists Sir William Ramsay and Morris M. Travers. A very common element throughout the universe, only 0.0018% of the earth’s atmosphere is neon. When ionized in a glass tube, neon emits a red light. This effect accounts for neon’s largest use in advertising lighting. Neon is also used to make high voltage indicators and is combined with helium to make helium-neon lasers. Liquid neon is used as a cryogenic refrigerant. Neon is highly inert and forms no known compounds, although there is some evidence that it could form a compound with flourine.
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。