Time-saving purification solutions for life-changing work
Empower your process with resins that can help bring your therapeutic product to market faster
Thermo Scientific purification solutions help customers by offering quality resins and technical guidance to support your downstream purification—from early-stage molecule development to large-scale manufacturing—in fewer steps and with high product yield.
Our commitment to your innovation and advancement in biotherapeutics
For more than three decades, Thermo Fisher Scientific has committed to advancing the downstream purification processes for valued customers like you. Uncover how persistent dedication to biotherapeutics innovation has yielded quality products and services, setting the stage for customers to advance the next generation of biotherapeutics.
From our first POROS products, to the launch of our CaptureSelect affinity resins and beyond, navigate through our milestones to learn about the technologies and when they were used to manufacture life-changing therapeutics.
POROS bead technology
1988
The POROS resin technology is first developed. The innovative bead features large through pores for improved process efficiency. The rigid, incompressible bead can withstand higher flow rates and is smaller than standard agarose beads, which leads to superior resolution properties.
POROS bead for purification
1992
The first POROS chromatography products launch: POROS HS and POROS HQ ion exchange chromatography resins. These cation and anion exchange resins are the first in their class featuring high dynamic binding capacity over a wide range of process conditions.
Camelid
1995
Researchers first identify Camelid single-domain antibodies. These unique antibodies form the basis of the CaptureSelect affinity technology, the first of its kind and currently the most comprehensive affinity ligand library and affinity resin portfolio in the world.
POROS resin bead for commercial manufacturing
1997
A POROS resin is used for the commercial manufacturing of a therapeutic antibody targeted against CD20, a surface antigen present in B-cells. This anti-CD20 antibody can be used to treat auto-immune diseases and certain types of cancer.
CaptureSelect affinity matrix bottle
1997
CaptureSelect Human Albumin Affinity Matrix is used for the capture of human serum albumin and albumin fusion proteins.
POROS resins for purification therapeutic
1998
POROS products are used in the commercial manufacture of a therapeutic antibody against RSV protein F, a treatment that protects high-risk babies from deadly Respiratory Syncytial Virus (RSV) infections.
virus molecule
2003
Thermo Scientific POROS HS cation exchange resin is used in the development of a human papillomavirus (HPV) virus-like particle vaccine. The vaccine is used to prevent cervical cancer in young women, caused by HPV infection.
POROS bead for purification
2004
POROS is used in the commercial manufacturing process of a therapeutic antibody against VEGF-A (vascular endothelial growth factor A), which functions as an angiogenesis inhibitor. The therapy is used as a supporting drug in various types of cancer and to treat a specific eye disease.
POROS cation exchange resin bead
2009
A major improvement on traditional cation exchangers, the newly launched Thermo Scientific POROS XS resin sets the standard for cation exchange chromatography. POROS XS is a strong anion exchange resin with increased capacity and high resolution.
CaptureSelect FSH Affinity Matrix
2013
First CaptureSelect resin was launched for the purification of therapeutic FSH (follicle-stimulating hormone). FSH plays an important role in the reproductive process.
monoclonal antibody
2013
A type of cancer immunotherapy uses a therapeutic monoclonal antibody that blocks the PD-1 receptor, a checkpoint inhibitor protein present on the surface of T- and B-cells.
Antibody diagram
2014
Thermo Fisher launches the antibody affinity resin portfolio, developed to overcome purification challenges associated with novel antibody formats. These resins are the alternative solution when Protein A doesn’t yield acceptable results.
Blood cells
2015
Recombinant FVIII, an innovative hemophilia biotherapeutic, approved using CaptureSelect technology in its manufacturing process.
POROS XS strong anion exchange resin
2015
Launched the Thermo Scientific POROS XQ resin—a next-generation, high-capacity, high-resolution, salt-tolerant strong anion exchange resin that allows loading to more than 140 mg/mL capacity in the presence of higher salt while delivering exceptional separation capability.
virus molecule
2015
By combining the two novel technologies, a unique purification solution for AAV vectors is launched. CaptureSelect AAV8 and AAV9 affinity resins address the purification challenges of AAV development, a vector broadly used in gene therapy.
hydrophobic interaction resins
2017
Launch of POROS hydrophobic interaction chromatography family. Covering a wide range of hydrophobicities, these high performance bioprocess resins are engineered for improved impurity removal during large-scale downstream purification of biomolecules.
virus particle for purification
2017
Introduction of POROS CaptureSelect AAVX Affinity Resin, a universal capture resin for AAV vector purification. The launch of POROS CaptureSelect AAVX is an extension of the AAV purification portfolio, engineered to purify a broad range of naturally occurring and synthetic AAV serotypes used for gene therapies—from AAV1 to AAV9, AAVrh10, and chimeric vectors.
Mosquitos that carry malaria
2019
CaptureSelect C-Tag affinity matrix is used in the downstream process of a protein-based malaria vaccine used in clinical trials.
DNA helix
2020
Launch of POROS Oligo (dT)25, designed for the purification and isolation of mRNA from in vitro transcription manufacturing processes. The resin effectively separates mRNA from components of the transcription reaction process, such as enzymes and plasmid DNA.
Blood cells
2020
A recombinant Factor VII product, used to treat a rare blood clotting disorder, is approved using CaptureSelect technology for manufacturing.
clusterin clear resin
2021
Launch of the first POROS CaptureSelect resin which can be used for the removal of Clusterin, a host cell protein that is difficult to remove.
Resin for T-cell subset isiolation
2021
Launch of CaptureSelect products featuring CD4 and CD8 ligands for the isolation of specific subsets of T-cells.
monoclonal antibody for purificiaton
2021
A high-performance Protein A resin, Thermo Scientific POROS MabCaptureC affinity resin is launched. Designed with high capacity and excellent alkaline stability, this resin offers efficient process-scale purification of monoclonal antibodies.
Mixed-mode resin
2024
To meet the demands of complex monoclonal antibody purification development and manufacturing, the Thermo Scientific POROS Caprylate Cation Exchange Chromatography Resin is released. This resin helps boost purification through the combined principles of both ion exchange and hydrophobic interactions.
Developed for the purification of engineered monoclonal antibody-based therapeutics, this selection of affinity chromatography resins can help you solve challenges in the downstream process of next-generation antibody therapeutics.
Camelid
1995
Researchers first identify Camelid single-domain antibodies. These unique antibodies form the basis of the CaptureSelect affinity technology, the first of its kind and currently the most comprehensive affinity ligand library and affinity resin portfolio in the world.
POROS resin bead for commercial manufacturing
1997
A POROS resin is used for the commercial manufacturing of a therapeutic antibody targeted against CD20, a surface antigen present in B-cells. This anti-CD20 antibody can be used to treat auto-immune diseases and certain types of cancer.
POROS bead for purification
2004
POROS is used in the commercial manufacturing process of a therapeutic antibody against VEGF-A (vascular endothelial growth factor A), which functions as an angiogenesis inhibitor. The therapy is used as a supporting drug in various types of cancer and to treat a specific eye disease.
monoclonal antibody
2013
A type of cancer immunotherapy uses a therapeutic monoclonal antibody that blocks the PD-1 receptor, a checkpoint inhibitor protein present on the surface of T- and B-cells.
Antibody diagram
2014
Thermo Fisher launches the antibody affinity resin portfolio, developed to overcome purification challenges associated with novel antibody formats. These resins are the alternative solution when Protein A doesn’t yield acceptable results.
monoclonal antibody for purificiaton
2021
A high-performance Protein A resin, Thermo Scientific POROS MabCaptureC affinity resin is launched. Designed with high capacity and excellent alkaline stability, this resin offers efficient process-scale purification of monoclonal antibodies.
Mixed-mode resin
2024
To meet the demands of complex monoclonal antibody purification development and manufacturing, the Thermo Scientific POROS Caprylate Cation Exchange Chromatography Resin is released. This resin helps boost purification through the combined principles of both ion exchange and hydrophobic interactions.
Explore related pages for these antibody purification solutions:
To support the purification of vaccines, we offer a broad variety of innovative purification solutions to improve the downstream processing of various vaccine modalities.
POROS resins for purification therapeutic
1998
POROS products are used in the commercial manufacture of a therapeutic antibody against RSV protein F, a treatment that protects high-risk babies from deadly Respiratory Syncytial Virus (RSV) infections.
virus molecule
2003
Thermo Scientific POROS HS cation exchange resin is used in the development of a human papillomavirus (HPV) virus-like particle vaccine. The vaccine is used to prevent cervical cancer in young women, caused by HPV infection.
Mosquitos that carry malaria
2019
CaptureSelect C-Tag affinity matrix is used in the downstream process of a protein-based malaria vaccine used in clinical trials.
To support the manufacturing of cell and gene therapies, we offer chromatography solution resins designed to improve the downstream purification of viral vectors.
virus molecule
2015
By combining the two novel technologies, a unique purification solution for AAV vectors is launched. CaptureSelect AAV8 and AAV9 affinity resins address the purification challenges of AAV development, a vector broadly used in gene therapy.
virus particle for purification
2017
Introduction of POROS CaptureSelect AAVX Affinity Resin, a universal capture resin for AAV vector purification. The launch of POROS CaptureSelect AAVX is an extension of the AAV purification portfolio, engineered to purify a broad range of naturally occurring and synthetic AAV serotypes used for gene therapies—from AAV1 to AAV9, AAVrh10, and chimeric vectors.
We have developed an exceptional portfolio of affinity resins to help you develop biosimilars, biobetters, and other types of recombinant proteins. Discover affinity resins that can be used for clinical and commercial production.
CaptureSelect affinity matrix bottle
1997
CaptureSelect Human Albumin Affinity Matrix is used for the capture of human serum albumin and albumin fusion proteins. These resins are the first-of-their-kind on the market.
CaptureSelect FSH Affinity Matrix
2013
First CaptureSelect resin was launched for the purification of therapeutic FSH (follicle-stimulating hormone). FSH plays an important role in the reproductive process.
Blood cells
2015
Recombinant FVIII, an innovative hemophilia biotherapeutic, approved using CaptureSelect technology in its manufacturing process.
Blood cells
2020
A recombinant Factor VII product, used to treat a rare blood clotting disorder, is approved using CaptureSelect technology for manufacturing.
clusterin clear resin
2021
Launch of the first POROS CaptureSelect resin which can be used for the removal of Clusterin, a host cell protein that is difficult to remove.
With the emergence of novel nucleic acid therapeutics, we have established high-performing chromatography resins to help you improve efficiency and productivity in the purification of mRNA and plasmid DNA.
DNA helix
2020
Launch of POROS Oligo (dT)25, designed for the purification and isolation of mRNA from in vitro transcription manufacturing processes. The resin effectively separates mRNA from components of the transcription reaction process, such as enzymes and plasmid DNA.
Beginning with the development of the POROS bead-based chromatography resins, Thermo Fisher Scientific’s purification solutions are designed to enhance process efficiency, resolution, and impurity removal in biomolecule purification. 
POROS bead technology
1988
The POROS resin technology is first developed. The innovative bead features large through pores for improved process efficiency. The rigid, incompressible bead can withstand higher flow rates and is smaller than standard agarose beads, which leads to superior resolution properties.
POROS bead for purification
1992
The first POROS chromatography products launch: POROS HS and POROS HQ ion exchange chromatography resins. These cation and anion exchange resins are the first in their class featuring high dynamic binding capacity over a wide range of process conditions.
POROS cation exchange resin bead
2009
A major improvement on traditional cation exchangers, the newly launched Thermo Scientific POROS XS resin sets the standard for cation exchange chromatography. POROS XS is a strong anion exchange resin with increased capacity and high resolution.
POROS XS strong anion exchange resin
2015
Launched the Thermo Scientific POROS XQ resin—a next-generation, high-capacity, high-resolution, salt-tolerant strong anion exchange resin that allows loading to more than 140 mg/mL capacity in the presence of higher salt while delivering exceptional separation capability.
hydrophobic interaction resins
2017
Launch of POROS hydrophobic interaction chromatography family. Covering a wide range of hydrophobicities, these high performance bioprocess resins are engineered for improved impurity removal during large-scale downstream purification of biomolecules.
Resin for T-cell subset isiolation
2021
Launch of CaptureSelect products featuring CD4 and CD8 ligands for the isolation of specific subsets of T-cells.
Explore related pages for these antibody purification solutions:
Related resources
Use our resin selection tool to help determine the right resin for your modality. 
Chromatography Learning Lab
Explore digital resources for purification resin application in the Chromatography Learning Lab.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.                                                                                                                                 
Connect with a Bioprocessing representative or request more info.                                          
Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.