Whether you are in the lab or out in the field, this aqueous, non-toxic Tissue Collection:RNA Stabilization Solution functions from ambient temperatures down to -20°C. Here is a snapshot of how researchers have used RNAlater® in exciting discoveries around the world.
Pancreas--Germany
Isolating intact RNA from pancreas can be difficult because of high levels of nucleases expressed in this tissue. In a study about circadian expression of clock genes, scientists used RNAlater to collect rat pancreata at three hour intervals for eventual quantitative RT-PCR analyses. These experiments provide growing evidence of circadian pacemakers in islets and begin to examine the role of dysrhythmic insulin secretion at the onset of diabetes mellitus. To learn more, see FEBS Lett (2004) 564: 91-96.
Blood--United States
RNAlater can be used to protect RNA in blood samples for any research or diagnostic test. One research group has shown that this may be critical for maintaining gene expression patterns in peripheral blood mononuclear cells (PBMCs). When PBMCs were resuspended in RNAlater within 30 minutes of collection, major changes in gene expression profiles related to ex vivo stress response pathways or to RNA degradation were reduced. To learn more, see Genes Immun (2004) 5: 347-353.
Environmental Bacteria--Netherlands Antilles
To study differential gene expression in bacterial communities associated with coral black band disease (BBD), scuba-diving scientists collected active BBD mat samples from infected coral surfaces and BBD mat samples that had been moved 0.3 to 1 m from the reef. Upon removal from the ocean, their samples were stabilized in RNAlater for eventual RNA-arbitrarily primed PCR (RAP-PCR) and RT-PCR analyses. These experiments are providing important clues to the global problem of BBD migration as well as the infection and destruction of coral reefs by BBD. To learn more, see Appl Environ Microbiol (2004) 70: 3687-94.
Environmental Virus--Laos People's Democratic Republic
To find ways of improving diagnostic testing of classical swine fever (hog cholera), researchers simulated collection and transport conditions in tropical ambient temperatures for virus-positive spleen samples in RNAlater or a glycerol/saline buffer. In both widely used diagnostic tests (RT-PCR and antigen-capture ELISA), viral detection was more sensitive in samples preserved with RNAlater. To learn more, see J Virol Methods (2004) 118: 33-7.
Ordering Information
Catalog # Name Size Price (USD) Qty
AM7020 RNAlater™ Stabilization Solution, 100 mL Each
206.65
Online Exclusive
215.00
Save 8.35 (4%)
AM7022 RNAlater™ Stabilization Solution, 50 x 1.5 mL Each
387.00
AM7021 RNAlater™ Stabilization Solution, 500 mL Each
660.00
AM7024 RNAlater™ Stabilization Solution, 1 x 250 mL Each
458.65
Online Exclusive
481.00
Save 22.35 (5%)
AM7023 RNAlater™ Stabilization Solution, 20 x 5 mL Each
388.65
Online Exclusive
407.00
Save 18.35 (5%)