Search
Materials Required
Protein
You may conjugate amine-reactive reagents with virtually any protein or peptide. The following protocol is optimized for IgG antibodies. You may scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results.
Reactive Dye
TFP, SDP, and succinimidyl esters are preferred for the conjugation to proteins because they form a very stable amide bond between the dye or hapten and the protein. Isothiocyanates are also commonly used, although the resulting thiourea product has been reported to deteriorate over time. Sulfonyl chlorides and acid halides are more reactive and may allow conjugation to aromatic amines. Sulfonyl chlorides form very stable sulfonamides that can survive complete protein hydrolysis, but since they are more difficult to work with, we do not recommend these for most routine conjugations with proteins.
Solvent
For the most part, reactive dyes and haptens are hydrophobic molecules and should be dissolved in high-quality, anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO). Caution: Do not use DMSO with sulfonyl chlorides, as it reacts with them.
Reaction Buffer
Amine-reactive reagents react with non-protonated aliphatic amine groups, including the amine terminus of proteins and the ε-amino group of lysines. The ε-amino group has a pKa of around 10.5; in order to maintain this amine group in the non-protonated form, the conjugation must take place in a buffer with slightly basic pH. It is important to avoid buffers that contain primary amines, such as Tris, as these will compete for conjugation with the amine-reactive compound. We recommend the following buffers for conjugation with aminereactive compounds with proteins:
You may achieve more specific labeling of the amine terminus using a buffer closer to neutral pH, as the pKa of the terminal amine is lower than that of the lysine ε-amino group.
Stop Reagent (Optional)
You may use 1.5 M hydroxylamine, pH 8.5, to terminate the reaction and to remove weakly bound probes. To prepare this reagent, dissolve hydroxylamine hydrochloride at 210 mg/mL in distilled water and adjust the pH to 8.5 with 5 M NaOH. Dilute the resulting 3 M solution with an equal volume of distilled water to get 1.5 M hydroxylamine, pH 8.5. Prepare this reagent freshly before use (see step 1.5 below).
Purification
You can easily separate a typical labeled antibody from free dye using a gel filtration column, such as Sephadex™ G-25, BioGel® P-30, or equivalent, equilibrated with the buffer of your choice. For much smaller or larger proteins, select a gel filtration media with a suitable molecular weight cutoff, or purify the conjugate by extensive dialysis (see step 2.1 below).
1. LABELING PROTOCOL
You may scale this procedure up or down, maintaining the same molar ratios of reagents. It is important to consider that the number and surface position of the amines will vary greatly among proteins and even among different IgGs, as will the reactivity of the dyes. We recommend that you try three different degrees of labeling whenever possible, using three different molar ratios of the reactive reagent to protein, and base future protocols on the amount of reagent that gives the most satisfactory results for your specific protein. Reviews by Brinkley and by Haugland provide comprehensive surveys of procedures and reagents for protein conjugate preparation.2,3
Note: Treatment with hydroxylamine at this stage is required to remove the trifluoroacetyl protecting groups of Rhodamine Green™ (Cat. no. R6112) succinimidyl ester. Hydroxylamine may also remove dye from unstable conjugates with tyrosine, serine, threonine and histidine.
2. SEPARATING THE CONJUGATE FROM THE UNREACTED LABELING REAGENT
We recommend using Sephadex™ G, BioGel® P, or equivalent gel filtration media with an appropriate molecular weight cutoff.
If you prefer to purify your conjugate by column chromatography, after elution, add bovine serum albumin (BSA) or any other stabilizer of choice to a final concentration of 1–10 mg/mL to prevent denaturation.
STORING THE PROTEIN CONJUGATE
Store the conjugates under the same conditions used for the parent protein. For storage in solution at 2–6°C, add sodiumazide (2 mM final concentration) as a preservative. Since azide is an inhibitor of horseradish peroxidase (HRP), substitute thimerosal as a preservative for conjugates that are derived from HRP or those that will be used for experiments in which HRP is present. It may be necessary to remove the preservatives prior to use to avoid inhibitory effects in applications in which conjugates are added to live cell specimens.
DETERMINING THE DEGREE OF LABELING
You may need to optimize the labeling efficiency to achieve the desired results in your application. You can determine the relative efficiency of a labeling reaction by measuring the abs
A = ε × path length × concentration
where є is the extinction coefficient in cm–1M–1. You also need to make a correction for the absorbance of the dye at 280 nm (see below). In the case of a biotinylated conjugate, you can measure the degree of labeling with HABA reagent as described in the protocol of our FluoReporter® Biotin-XX Protein Labeling Kit and elsewhere.4
![]() |
![]() |
![]() |
where MW = the molecular weight of the protein, εdye = the extinction coefficient of the dye at its absorbance maximum, and the protein concentration is in mg/mL.
Materials Required
Oligonucleotide
The oligonucleotide must be synthesized with an amine group on the 5′ end. The following protocol has been optimized for use with 100 μg of oligonucleotide.
Reactive Dye
Succinimidyl esters are preferred for the conjugation to oligonucleotides because they are easy to use, and form a very stable amide bond between the dye or hapten and the amine-modified oligonucleotide. For DNA sequencing applications, the BODIPY® dyes are useful because they are isomerically pure and cause little perturbation to the mobility of DNA fragments.5 We also offer 5-FAM, 6-TAMRA, 6-ROX, and 6-JOE, the traditional fluorophores used in oligonucleotide labeling and automated DNA sequencing applications.6-9 The Alexa Fluor® series of dyes provides very bright, photostable fluorescence; however, because the fluorescence of the Alexa Fluor® 633 dye appears to be quenched by nucleic acids, we do not recommend this dye for labeling oligonucleotides.
Solvent
For the most part, reactive dyes and haptens are hydrophobic molecules and should be dissolved in high-quality, anhydrous dimethylsulfoxide (DMSO) before reaction with aminemodified oligonucleotides.
Reaction Buffer
Amine-reactive reagents react with the non-protonated amine group on the modified oligonucleotide. To maintain this amine group in the non-protonated form, the conjugation must take place in a buffer with slightly basic pH. For optimal results, we recommend using a tetraborate buffer at pH 8.5, rather than the bicarbonate buffers recommended for protein conjugations. Avoid buffers that contain primary amines, such as Tris, as these compete for conjugation with the amine-reactive compound.
PURIFYING THE AMINE-MODIFIED OLIGONUCLEOTIDE
To ensure that the oligonucleotide is free of interfering compounds, especially amines, such as triethylamine or Tris, and ammonium salts, we strongly recommend extracting and precipitating the sample prior to initiating the labeling reaction. We suggest the following protocol for 0.1–1 mg oligonucleotide (3–30 A260 units).
4.1 Dissolve the oligonucleotide in 100 μL of deionized water, and extract three times with an equal volume of chloroform.
4.2 Precipitate the oligonucleotide by adding one-tenth volume (10 μL) of 3 M NaCl and two and a half volumes (250 μL) of cold absolute ethanol. Mix well and incubate at ≤–20°C for 30 minutes.
4.3 Centrifuge the solution in a microcentrifuge at ~12,000 × g for 30 minutes.
4.4 Carefully remove the supernatant, rinse the pellet once or twice with cold 70% ethanol, and dry under vacuum.
4.5 Dissolve the dry pellet in deionized water to achieve a final concentration of 25 μg/μL (4.2 mM for an 18-mer). You may store this amine-modified oligonucleotide stock solution frozen at ≤–20°C.
LABELING PROTOCOL
The protocol has been optimized for labeling 100 μg of an 5′-amine– modified oligonucleotide, 18 to 24 bases in length. You may label slightly shorter or longer oligonucleotides using the same procedure; however, adjustments to the protocol may be necessary for significantly shorter or longer oligonucleotides. You may scale the reaction up or down as long as you do not change the concentration of each component. The procedure has not been tested with oligonucleotides containing more than one amine. Following the labeling reaction, you may purify the conjugate from the reaction mixture using preparative gel electrophoresis or reverse-phase HPLC.
5.1 Prepare 0.1 M sodium tetraborate, pH 8.5 labeling buffer.
a. Make a 0.1 M sodium tetraborate buffer by dissolving 0.038 g of sodium tetraborate decahydrate for every mL of water.
b. Adjust pH with HCl to 8.5. Prepare this labeling buffer as close as possible to the time of labeling. Alternatively, you may divide the buffer into small aliquots, and freeze immediately for long-term storage.
Note: Exposure of this solution to air for a long time results in carbon dioxide absorption, which changes the pH of the buffer.
5.2 Dissolve 250 μg of the amine-reactive compound in 14 μL DMSO. For dinitrophenyl–X (DNP–X) succinimidyl ester, use 160 μg for 100 μg of oligonucleotide. For fluorescein, tetramethylrhodamine, Marina Blue®, FAM, or TAMRA succinimidyl ester, use 200 μg for 100 μg of oligonucleotide.
a. Allow amine-reactive compound to equilibrate to room temperature before opening the vial. Do not heat.
b. Dissolve the material by pipetting up and down, washing the sides of the vial. Texas Red® compounds tend to precipitate easily, and may require longer times to completely dissolve.
Note: It is important that you prepare the amine-reactive label freshly for each labeling reaction as reactive compounds are not stable in solution.
5.3 To the vial containing the reactive label in DMSO, add:
The reaction mixture may have a grainy appearance, but this does not adversely affect the conjugation. We strongly advise against attempting to improve the solubility of the label, because modifying the composition of the mixture can drastically reduce the labeling efficiency. You may scale the reaction up or down as long as the concentration of each component is not changed. Do not add more dye than recommended, as excess dye will not improve the labeling efficiency, and may make the purification more difficult.
5.4 Incubate the reaction for at least six hours (or overnight if more convenient) at room temperature.
a. Place the vial on a shaker oscillating at low speed, gently vortex to mix, or tap the vial every half hour for the first two hours to ensure that the reaction remains well mixed. Do not mix violently, as material may be left on the sides of the vial.
b. After six hours, 50–90% of the amine-modified oligonucleotide molecules will be labeled. Allowing the incubation to proceed overnight does not necessarily result in a greater labeling efficiency.
ETHANOL PRECIPITATING THE LABELED OLIGONUCLEOTIDE
Following the reaction, the labeling mixture contains labeled oligonucleotide, unlabeled oligonucleotide, and unincorporated dye (or biotin or DNP). You can purify the labeled oligonucleotide from the reaction mixture using preparative gel electrophoresis or reverse-phase HPLC. Regardless of the purification method, we recommend ethanol precipitation as the first step.
6.1 Add one-tenth volume of 3 M NaCl and two and a half volumes of cold absolute ethanol to the reaction vial.
6.2 Mix well and incubate at ≤–20°C for 30 minutes.
6.3 Centrifuge the solution in a microcentrifuge at ~12,000 × g for a full 30 minutes. Loss of sample may occur if the centrifugation is not long enough.
6.4 Carefully remove the supernatant, rinse the pellet once or twice with cold 70% ethanol, and dry briefly. If the labeled oligonucleotide becomes completely dry, it will be difficult to redissolve.
PURIFYING THE LABELED OLIGONUCLEOTIDE BY HPLC
You can purify labeled oligonucleotides by reverse-phase HPLC using a standard analytical (4.6 × 250 mm) C8 column.
7.1 Dissolve the pellet from the ethanol precipitation (step 5.4) in 0.1 M TEAA (triethylammonium acetate).
7.2 Load the dissolved pellet onto the column in 0.1 M TEAA and run a linear 5–65% acetonitrile gradient over 30 minutes. This gradient is a 2% increase in acetonitrile per minute.
PURIFYING THE LABELED OLIGONUCLEOTIDE BY GEL ELECTROPHORESIS
8.1 To purify the labeled oligonucleotide by gel electrophoresis, pour a 0.5 mm–thick polyacrylamide slab gel. For oligonucleotides less than 25 bases in length, use 19% acrylamide, for oligonucleotides 25–40 bases, 15% acrylamide, and for oligonucleotides 40–100 bases, 12% acrylamide.
8.2 Resuspend the pellet from ethanol precipitation in 200 μL of 50% formamide, and incubate at 55°C for 5 minutes to disrupt any secondary structure.
8.3 Load the warmed oligonucleotide onto the gel (you may need to use several wells) and load an adjacent well with 50% formamide plus 0.05% bromophenol blue.
8.4 Run the gel until the bromophenol blue indicator dye is two-thirds of the way down the gel. The bromophenol blue will migrate at approximately the same rate as the oligonucleotide.
8.5 Remove the gel from the glass plates, place on Saran Wrap, and lay it on a fluorescent TLC plate. Locate the labeled and unlabeled oligonucleotides by illuminating with a handheld UV source. Fluorophore-labeled oligonucleotides show fluorescence when illuminated with UV light.
8.6 Cut out the band containing the labeled oligonucleotide and purify by the “crush-and-soak” method or other suitable method. For more details, refer to Sambrook J., Fritsch E.F., and Maniatis, T., Molecular Cloning: A Laboratory Manual, Second Edition, Cold Spring Harbor Laboratory (1989).