Search Thermo Fisher Scientific
The XSQ Tools project contains a set of utilities for XSQ file format conversions. Currently, this includes conversion from CSFASTA+QUAL to XSQ, conversion from XSQ to CSFASTA+QUAL, conversion to FASTQ, and splitting an XSQ file by its constituent libraries.
Please note that downloading and using this software indicates acceptance of the EULA terms.
Updates & Patches | |||||
![]() | Linux | Download | XSQ Software (Registration required) | ||
Documentation | |||||
![]() | Download | XSQ Tools User Guide | |||
![]() | Download | The Extensible Sequence (XSQ) File Format Specification v1.0.1 | |||
![]() | Download | The Extensible Sequence (XSQ) Webinar as presented on February 15, 2011 | |||
![]() | Download | Third Party Licenses | |||
Support |
| This file format is being released as "unsupported software" by Applied Biosystems. Please email your FBS for guidance. |