Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios with your Sanger sequencing experiments.
View the relevant questions below:
Beginning your experiment?
Visit our
For data run on capillary electrophoresis platforms, the troubleshooting process consists of the following steps:
Control DNA template (pGEM) and -21 M13 Primer are provided in the BigDye™ Terminator kit and can help you determine whether failed reactions are caused by poor template quality or sequencing reaction failure. See the table below for setting up the pGEM control.
Component
| Quantity per reaction
|
BigDye™ Terminator 3.1 Ready Reaction Mix
| 8 μL
|
–21 M13 Control Primer (0.8 pmol/μL)
| 4 μL
|
pGEM (200 ng/μL)
| 1-2 μL
|
Water
| Adjust based on pGEM volume. i.e., 7 or 6 μL to bring final reaction volume to 20 μL
|
Total volume
| 20 μL
|
Troubleshooting can be a time-consuming process but a methodical approach can save both time and money in finding the root problem and addressing it correctly. The Q/As below are some of the more common DNA Sequencing issues. However, additional troubleshooting can be found in the DNA Sequencing by Capillary Electrophoresis Guide.
Shoulders on all the peaks can be caused by the following:
If either G or C shouldering is occurring and the A and T peaks do not exhibit shouldering, it is possible that dye degradation is occurring. The G or C dye degradation may be due to photobleaching, oxidation, arcing event, or changes in pH. To prevent dye degradation, ensure the following:
There are many reasons for a noisy baseline, the most common being the following:
Peaks observed within the first 100 bp can be due to excess dye terminators (ddNTPs) remaining in the sample, also known as dye blobs. These dye blobs can occur in several positions within the first 100 bp and are typically seen as broad C, G, or T peaks and can impact basecalling. Additional optimization of the DNA sequencing cleanup will be needed. Please see our suggestions below:
Flat peaks are typically an indication of data with too high of a signal, also known as offscale data. Please check the raw data of the sample file and determine the signal intensity on the Y-axis. The maximum signal thresholds for raw data are 32,000 rfu for the Applied Biosystems™ 3730/3730xl DNA Analyzers and 3500/3500xL Genetic Analyzers, and 8,000 rfu for the Applied Biosystems™ 310 Genetic Analyzer and Applied Biosystems™ 3130/3130xl Genetic Analyzers.
There are several options for reducing the signal, please see our suggestions below:
Offscale sequencing data can be prevented by optimizing the amount of template and primer in the reaction. The recommended amount of primer is 3.2 pmol and the amount of template is dependent on the template type:
DNA template
| Quantity used with most DNA sequencing purification protocols
| Quantity if using BigDye™ XTerminator™ Purification Kit*
|
PCR product:
|
|
|
100–200 bp
| 1–3 ng
| 0.5–3 ng
|
200–500 bp
| 3–10 ng
| 1–10 ng
|
500–1000 bp
| 5–20 ng
| 2–20 ng
|
1000–2000 bp
| 10–40 ng
| 5–40 ng
|
>2000 bp
| 20–50 ng
| 20–50 ng
|
Other template:
|
|
|
Single-stranded DNA
| 25–50 ng
| 10–50 ng
|
Double-stranded DNA
| 150–300 ng
| 50–300 ng
|
Cosmid, BAC
| 0.5–1.0 μg
| 0.2–1.0 μg
|
Bacterial genomic DNA
| 2–3 μg
| 1–3 μg
|
* The BigDye™ XTerminator™ Purification Kit is intended to purify the DNA sequencing reaction after thermal cycling by sequestering cycle-sequencing reaction components such as salt ions, unincorporated dye terminators, and dNTPs, to prevent their co-injection with dye-labeled extension products into a capillary electrophoresis DNA analyzer.
If, after using the recommended amount of template and primer, offscale data is still present, it may be necessary to further decrease the amount of template used in the sequencing reaction.
Our capillary electrophoresis systems allow the editing of the injection time. The injection time can be reduced and the sample can be re-injected. Depending on the signal intensity, the sample may also require serial dilution to reduce the signal. Please refer to the appropriate instrument user guide for instructions on editing the injection time.
It is common to observe noisy data after a homopolymer repeat due to stutter. Stutter can occur during the PCR amplification or cycle sequencing and is due to polymerase slippage. If the stutter occurs during PCR, the use of anchored primer may help in the sequencing. Some customers have found that they can get past poly(A) regions using a mixture of oligo dT18 primers with either a C, A, or G as the 3’ terminal dinucleotide or 2-base anchors. Sequencing in both directions can also assist with obtaining sequencing before and after the homopolymer repeat.
Low signal intensity can be caused by many factors including thermal cycler malfunction (in the case of an entire plate failure), poor cleanup, and insufficient sequencing template quantity/quality.
Control DNA template (pGEM) and -21 M13 Primer are provided in the BigDye™ Terminator kit and can help you determine whether failed reactions are caused by poor template quality or sequencing reaction failure. See the table below for setting up the pGEM control.
Component
| Quantity per reaction
|
BigDye™ Terminator 3.1 Ready Reaction Mix
| 8 μL
|
–21 M13 Control Primer (0.8 pmol/μL)
| 4 μL
|
pGEM (200 ng/μL)
| 1-2 μL
|
Water
| Adjust based on pGEM volume. i.e., 7 or 6 μL to bring final reaction volume to 20 μL
|
Total volume
| 20 μL
|
Sequence composition, such as hairpins or secondary structure, may be preventing the DNA polymerase from sequencing through the region. Redesigning the primers around the region may help. Some customers have reported that the use of additives such as 5-10% DMSO, betaine, or glycerol, or increasing the initial incubation from 1-10 mins may help denature the template, but we have not tested these in-house.
Some customers have reported that the use of additives such as 5-10% DMSO, betaine, or glycerol or increasing the initial incubation from 1-10 mins may help denature the template, but we have not tested these in-house.
Here are possible causes and solutions for a missing C peak:
If the raw data shows good signals and a clean baseline, the issue is most likely the setting in the Sequencing Analysis Software. Please check the Analysis Protocol used to analyze the data. This can be done in Sequencing Analysis. Go to the Analysis menu > select Analysis Protocol Manager > highlight the Analysis Protocol in use > Select Edit > go to the Basecalling Tab and select ‘At PCR Stop.’ Then click OK. Apply the edited Analysis Protocol to the sample and re-analyze. Ideally a minimum of 100 bp is recommended to ensure successful basecalling.
Lack of signal can be determined by looking at the scale of signal produced in the raw data view. Lack of signal can be caused by many factors which include problems in the sequencing reaction (template quantity/quality), thermal cycler malfunction (plate failure) and capillary electrophoresis failure (failing laser, air bubbles in lines, etc.):
The BigDye™ XTerminator™ Solution is expected to perform to specification if left out for no more than one 24-hour period. We cannot guarantee performance for reagent left out for a longer period of time.
Yes. The precipitate should be resuspended by warming the SAM™ Solution (remember, never heat the BigDye™ XTerminator™ Solution) to 30-37 degrees C with gentle vortexing. Bubbles will form on the surface of the SAM™ Solution when vortexed. These will dissipate within a few minutes. Allow the SAM™ Solution to cool to ambient temperature before use. Do not use it while warm as using warm SAM™ Solution will produce poor results.
Dye blobs in the sequencing data can be caused by the following factors:
Please see page 28 of the BigDye™ Xterminator™ Purification Kit manual for other troubleshooting tips.
Some possible reasons for reduced signal for smaller fragments may be:
Please see page 28 of the BigDye™ Xterminator™ Purification Kit manual for other troubleshooting tips.
A possible reason for no signal when using the BigDye™ XTerminator™ Purification Kit is that the capillaries are going into the BigDye™ XTerminator™ Solution. To alleviate this issue, please check the run module that is being used on the Genetic Analyzer. The template should have “BDX” in the run module name. If it does not, it means that the capillaries are going into the BigDye™ XTerminator™ Solution, and this will prevent the injection of the sample into the capillaries.
Some possible reasons for low signal when using the BigDye™ XTerminator™ Purification Kit are:
This is most likely due to incomplete conversion of the DNA template. Please see below for possible reasons why conversion may be incomplete:
There are several reasons why conversion may be incomplete:
If the DNA template was bisulfite converted successfully and the PCR amplification was successful, the issue may be due to long stretches of T, due to the C>T conversion, resulting in slippage and stutter peaks. Additionally, there may be secondary sequences or primer dimer sequences interfering with the sequencing reaction. The presence of these types of shorter amplicons can contribute to offscale signals at the beginning of the sequence trace, and depletion of the sequencing components, which causes rapid drop-off of signal intensity for the remaining sequence. If sequencing the PCR amplicon directly, the following list of recommendation may help with improving the sequencing reaction:
For Research Use Only. Not for use in diagnostic procedures.