Search
Saccharomyces cerevisiae cells were stained sequentially with the red-fluorescent rhodamine B hexyl ester (R 6, Cat. No. R648MP), which selectively labels yeast mitochondria under these conditions, and the green-fluorescent yeast vacuole membrane marker MDY-64 (Cat. No. Y7536). These probes are also provided in our Yeast Mitochondrial Stain Sampler Kit (Cat. No. Y7530) and Yeast Vacuole Marker Sampler Kit (Cat. No. Y7531), respectively. Stained yeast were photographed in a single exposure through an Omega Optical triple-bandpass filter set.
Staining with MDY-64. Go ›
Staining with Arg-CMAC. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›