Qdot® Nanocrystal Primary Antibody and Streptavidin Conjugates
Use the guide below to help you find the right Qdot® product for your application.
Description | Clone | Isotype | Qdot 565 | Qdot 605 | |
Qdot 655 | Qdot 705 | Qdot 800 |
---|---|---|---|---|---|---|---|---|
Human CD2 |
S5.5 | Mouse IgG2a |
|
Q10172 | |
|
|
|
Human CD3 | UCHT1 | Mouse IgG2a | Q10054 | |
|
|||
Human CD3 | S4.1 | Mouse IgG2a | Q10012 | |
|
|||
Human CD4 | S3.5 | Mouse IgG2a | Q10008 | Q10007 | Q10060 | |||
Human CD8 | 3B5 | Mouse IgG2a | Q10152 | Q10009 | Q10055 | Q10059 | ||
Human CD10 | MEM-78 | Mouse IgG1 | Q10153 | Q10154 | ||||
Human CD14 | TüK4 | Mouse IgG2a | Q10013 | Q10056 | Q10064 | |||
Human CD19 |
SJ25-C1 | Mouse IgG1 |
|
Q10306 | |
Q10179 | |
|
Human CD20 | HI47 | Mouse IgG3 | Q10305 | |||||
Human CD27 | CLB-27/1 | Mouse IgG2a | Q10065 | Q10066 | ||||
Human CD38 | HIT2 | Mouse IgG1 | Q10053 | Q10057 | ||||
Human CD45 | HI30 | Mouse IgG1 | Q10051 | Q10155 | Q10062 | Q10156 | ||
Human CD45RA | MEM-56 | Mouse IgG2b | Q10047 | Q10069 | ||||
Human CD56 | MEM-188 | Mouse IgG2b | |
Q10307 | |
|
|
|
Human HLA-DR | TÜ36 | Mouse IgG2b | Q10052 | Q10063 | ||||
Mouse CD3 | 145-2C11 | Hamster IgG | Q10090 | |||||
Mouse CD4 | RM4-5 | Rat IgG2a | |
Q10092 | |
|
|
|
Mouse CD19 |
6D5 |
Rat IgG2a |
|
|
Q10379 | |||
Mouse CD45R (B220) | RA3-6B2 | Rat IgG2a | |
Q10176 | |
|||
Mouse IgG1 | NA | Mouse IgG1 | Q10073 | |
|
Q10298 | ||
Mouse IgG2a | NA | Mouse IgG2a | Q10014 | Q10015 | Q10076 | Q10075 | ||
Mouse IgG2b | NA | Mouse IgG2b | Q10074 | |
||||
Rat IgG2a | NA | Rat IgG2a | |
|
|
|
|
|
Streptavidin | NA | NA | Q10131MP | Q10101MP | |
Q10121MP | Q10161MP | Q10171MP |
- All antibodies listed in this table are for Research Use Only.
- All antibodies and isotype controls listed available as 0.1 ml, 100 tests minimum size
- Streptavidin conjugates are available as 200µl or 500 µl (Q10161MP) sizes
Spectral properties and recommended filters for Qdot nanocrystals
Qdot Nanocrystal | Excitation (nm)* | Peak Emission | Recommended Filter |
---|---|---|---|
Qdot 565 | 405 (488) | 565 | 575/26 |
Qdot 605 | 405 (488) | 605 | 605/20 |
Qdot 625 | 405 (488) | 625 | 625/20 |
Qdot 655 | 405 (488)† | 655 | 655/20 |
Qdot 705 | 405 (488)† | 705 | 720/20 |
Qdot 800 | 405 (488)† | 800 | 787/42 |
† Can also be excited by 633 nm lasers
サポートノート
注)全てのQdot®及びQtracker® には毒物及び劇物取締法の毒物に該当するセレン化合物が含まれており、
また廃棄物処理法等による規制対象であるカドミウムも含まれています。取り扱い、保管、廃棄等にはご注意下さい。
また廃棄物処理法等による規制対象であるカドミウムも含まれています。取り扱い、保管、廃棄等にはご注意下さい。
For Research Use Only. Not for use in diagnostic procedures.