Sodium pyruvate
Sodium pyruvate is supplied as a liquid supplement for cell culture media at 100 mM (100X) and a final concentration of 11,004 mg/L.
Mammalian cell culture with our classical cell culture media
For all your additional mammalian cell culture needs checkout out line of GIBCO® brand classical media products: