E-Gel CloneWell II gels are compatible with the
The smartest way to gel-purify your DNA
  • Gel-purify your DNA in 3 simple steps
  • Get improved cloning efficiencies
  • View bands in real time and minimize DNA damage
  • Collect multiple DNA bands from the same gel lane
Order nowRequest a demo
Download the E-Gel CloneWell II flyer
E-Gel CloneWell II gels are compatible with the E-Gel Power Snap System
Gel-purify your DNA in 3 simple steps
E-Gel CloneWell II agarose gels are double-comb gels with a twist. Load your sample into the top row and electrophorese until your band migrates into the bottom row (Figure 1). Then simply pipet out your purified DNA band and you’re ready to clone. That’s it. No additional purification kits or steps are required. Use the Invitrogen E-Gel Power Snap System, a compact, self-contained device with a built-in power supply and blue-light transilluminator, to run and visualize E-Gel CloneWell II agarose gels.
Three easy steps for separation and isolation of DNA bands with E-Gel CloneWell II agarose gels
Figure 1. Three easy steps for separation and isolation of DNA bands with E-Gel CloneWell II agarose gels. Samples are loaded into the top row of wells, bands separate during the gel run, and individual bands are collected from the bottom row as they enter those wells. Reverse-run functionality on the E-Gel Power Snap System lets you capture bands of interest even if you miss the bands when they pass through the collection wells.
Get improved cloning efficiencies
Exposure of your DNA sample to UV light during visualization may lead to DNA damage and reduced cloning efficiencies. Using E-Gel CloneWell II agarose gels with the E-Gel Power Snap System eliminates UV damage and improves cloning efficiency compared to conventional methods. Results obtained using the TOPO TA Cloning Kit after E-Gel CloneWell II gel purification, compared to a conventional method, are shown in Figure 2.
 Improved cloning efficiency using E-Gel CloneWell agarose gels with blue-light transillumination
Figure 2. Improved cloning efficiency using E-Gel CloneWell II agarose gels with blue-light transillumination. A PCR reaction containing an 850 bp amplicon was separated on either an E-Gel CloneWell agarose gel or a traditional agarose gel containing ethidium bromide. The DNA band retrieved from the E-Gel CloneWell agarose gel was visualized using blue-light transillumination. DNA separated on the traditional gel was viewed using UV light and isolated by first cutting a gel slice with a razor blade and then using a commercially available gel extraction kit. In both cases, the exposure of the DNA to the light source was 15, 30, or 60 sec. Both fragment samples were cloned using the TOPO TA Cloning Kit (Cat. No. K460040) and transformed into Invitrogen MultiShot TOP10 chemically competent cells (Cat. No. C40005). Shown are average numbers of colony-forming units (CFU) obtained for each exposure time and cloning method.
Collect multiple DNA bands from the same gel lane
With E-Gel CloneWell II agarose gels, you can retrieve multiple DNA bands from the same sample/lane, thus saving materials and time. Avoid the hassle of cutting out gel bands and using multiple columns for further gel extraction. With E-Gel CloneWell II agarose gels, just retrieve the bands one at a time as they migrate into the collection well. No additional purification is required.
View bands in real time and minimize DNA damage
The E-Gel Power Snap System allows real-time viewing of DNA band migration through E-Gel agarose gels containing SYBR Safe DNA Gel Stain. Unlike UV light, the blue light used in this system causes minimal DNA damage (Figure 3).
Figure 3. Use of SYBR Safe DNA Gel Stain and a blue-light transilluminator causes minimal DNA damage. Equivalent fractions of supercoiled DNA stained with SYBR Safe DNA Gel Stain or ethidium bromide were exposed to blue light (Safe Imager 2.0 Blue-Light Transilluminator) or UV light, respectively, for defined periods of time and evaluated by agarose gel electrophoresis. A slower-migrating species is indicative of a linear or relaxed circular vector that results from DNA nicking or strand breaks.
Gel-purify your DNA in 3 simple steps
E-Gel CloneWell II agarose gels are double-comb gels with a twist. Load your sample into the top row and electrophorese until your band migrates into the bottom row (Figure 1). Then simply pipet out your purified DNA band and you’re ready to clone. That’s it. No additional purification kits or steps are required. Use the Invitrogen E-Gel Power Snap System, a compact, self-contained device with a built-in power supply and blue-light transilluminator, to run and visualize E-Gel CloneWell II agarose gels.
Three easy steps for separation and isolation of DNA bands with E-Gel CloneWell II agarose gels
Figure 1. Three easy steps for separation and isolation of DNA bands with E-Gel CloneWell II agarose gels. Samples are loaded into the top row of wells, bands separate during the gel run, and individual bands are collected from the bottom row as they enter those wells. Reverse-run functionality on the E-Gel Power Snap System lets you capture bands of interest even if you miss the bands when they pass through the collection wells.
Get improved cloning efficiencies
Exposure of your DNA sample to UV light during visualization may lead to DNA damage and reduced cloning efficiencies. Using E-Gel CloneWell II agarose gels with the E-Gel Power Snap System eliminates UV damage and improves cloning efficiency compared to conventional methods. Results obtained using the TOPO TA Cloning Kit after E-Gel CloneWell II gel purification, compared to a conventional method, are shown in Figure 2.
 Improved cloning efficiency using E-Gel CloneWell agarose gels with blue-light transillumination
Figure 2. Improved cloning efficiency using E-Gel CloneWell II agarose gels with blue-light transillumination. A PCR reaction containing an 850 bp amplicon was separated on either an E-Gel CloneWell agarose gel or a traditional agarose gel containing ethidium bromide. The DNA band retrieved from the E-Gel CloneWell agarose gel was visualized using blue-light transillumination. DNA separated on the traditional gel was viewed using UV light and isolated by first cutting a gel slice with a razor blade and then using a commercially available gel extraction kit. In both cases, the exposure of the DNA to the light source was 15, 30, or 60 sec. Both fragment samples were cloned using the TOPO TA Cloning Kit (Cat. No. K460040) and transformed into Invitrogen MultiShot TOP10 chemically competent cells (Cat. No. C40005). Shown are average numbers of colony-forming units (CFU) obtained for each exposure time and cloning method.
Collect multiple DNA bands from the same gel lane
With E-Gel CloneWell II agarose gels, you can retrieve multiple DNA bands from the same sample/lane, thus saving materials and time. Avoid the hassle of cutting out gel bands and using multiple columns for further gel extraction. With E-Gel CloneWell II agarose gels, just retrieve the bands one at a time as they migrate into the collection well. No additional purification is required.
View bands in real time and minimize DNA damage
The E-Gel Power Snap System allows real-time viewing of DNA band migration through E-Gel agarose gels containing SYBR Safe DNA Gel Stain. Unlike UV light, the blue light used in this system causes minimal DNA damage (Figure 3).
Figure 3. Use of SYBR Safe DNA Gel Stain and a blue-light transilluminator causes minimal DNA damage. Equivalent fractions of supercoiled DNA stained with SYBR Safe DNA Gel Stain or ethidium bromide were exposed to blue light (Safe Imager 2.0 Blue-Light Transilluminator) or UV light, respectively, for defined periods of time and evaluated by agarose gel electrophoresis. A slower-migrating species is indicative of a linear or relaxed circular vector that results from DNA nicking or strand breaks.
Resources
Have questions about the E-Gel system or new to molecular biology applications? Access literature, selection guides, webinars, and additional educational resources below for more information.
Literature
Read our brochure, white papers, technical notes, and white papers to facilitate your experiments.
Videos
Watch fun and educational videos on molecular biology techniques, how-tos, and tips and tricks.
Webinars
View upcoming and recorded webinars for in-depth understanding of molecular biology techniques and resources.
Selection guide
Find our E-Gel selection table that will guide you in choosing the best products for your experiments.
Product support centers
Browse the support centers by application for technical support and troubleshooting help.
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications