More than 1,000 protein-specific ELISA kits
Ready-to-use Invitrogen ELISA kits are optimized for convenient and sensitive quantitation of a wide range of targets in various sample types, including human, mouse, rat IgG, cytokines, phosphorylated proteins, beta amyloids, extracellular and intracellular targets. Pre-coated and coat-it-yourself sandwich ELISA kits include capture and detection matched antibodies, standards, buffers, diluents, and detection reagents. Specialty ELISA formats and configurations are also available.
We offer several varieties each of coated and uncoated ELISA kits, including those based on Invitrogen Instant ELISA Technology, validated for particular levels of sensitivity, formatted as sandwich and competitive ELISAs, or optimized for a various sample types. Our portfolio of ELISA kits now includes products previously sold under the Affymetrix eBioscience brand, now part of Thermo Fisher Scientific.
The new, combined Invitrogen ELISA kit product family encompasses a greater range of available analytes, species, and formats. Especially noteworthy additions are the Instant ELISA kits and expanded line of immunoglobulin and isotyping kits.
Learn more about ELISA kit formats
Complete and validated.
These kits include antibodies, standards, detection reagents, and 96-well plates that are precoated with the capture antibody.
Affordable and convenient.
These kits contain matched antibody pairs, standards and detection reagents but require users to coat the capture antibody themselves.
Customize to your needs.
Develop, test, and validate your own direct or sandwich ELISA by selecting your own antibodies, reagents, plates, and other assay components.
Coated ELISA Kits › Uncoated ELISA Kits ›
Antibodies Pairs ›
ELISA Reagents & Buffers ›
Microplates & Plasticware ›
Featured categories of ELISA kits
Cytokine, Chemokine, and Growth Factor ELISA Kits
Over 700 cytokine and extracellular ELISA kits covering broad range of targets in several species and various sample types.
Immunoglobulin ELISA Kits
Antibody isotyping ELISA kits to identify and measure IgG, IgA, IgE, and IgM subclasses and isotypes from human, mouse and rat species.
Neurobiology ELISA Kits
Tau, amyloid beta, and other target-specific ELISA kits designed for neuroscience researchers and those studying neurological disorders.
Phospho-Specific ELISA Kits
Phospho-serine, threonine, and tyrosine ELISA and InstantOne kits, to measure intracellular protein phosphorylation, signaling pathways, and activation.
Learn more about our methods of ELISA Validation and Quality Testing