Neurobiology ELISA Kits
ELISA kits for neuroscience research
Invitrogen ELISA kits for neurobiology are high-quality ELISA kits, designed for researchers studying neurological disorders or doing neuroscience-related research. These “neuroELISA” kits are designed to deliver accurate, sensitive, and fast Aβ (β-amyloid or amyloid beta), tau, and α-synuclein protein quantitation in sample types such as cerebrospinal fluid (CSF), cell culture supernatants, primary neurons, and brain tissue samples.
Select neurobiology ELISA kits from among all target-specific ELISA kits using the following selection guide.
Find ELISA kits by target
Features of neuro-specific Invitrogen ELISA kits
  • Confidence in results—consistent, accurate, and sensitive measurements
  • Rigorous quality control—kits must meet quality-controlled specifications for sensitivity, dynamic range, precision, specificity, recovery, and lot-to-lot consistency
  • Ease of use—kits include pre-coated plates (8-well strip-based format), capture/detection antibodies, standards, buffers and accessory reagents
Study proteins implicated in neurodegenerative disorders
ELISA enables specific, quantitative measurements of protein from a variety of biological sources including serum, plasma, and CSF (cerebrospinal fluid) from a variety of species. We offer ELISA kits for study of several proteins implicated in neurodegenerative disorders, such as:
  • Amyloid-beta (Aβ, beta amyloid or Abeta), which is crucially involved in Alzheimer's disease as the main component of the amyloid plaques found in the brains of Alzheimer’s patients
  • Tau, a protein which stabilizes microtubules and which, when not functioning properly, is implicated in pathologies and dementias of the nervous system
  • Alpha synuclein (α-synuclein) which is thought to play an important role in maintaining a supply of synaptic vesicles in presynaptic terminals, and may also help regulate the release of dopamine
Performance data for the Invitrogen Human Tau (Total) ELISA Kit (Cat. No. KHB0041). Left panel: Typical 7-point standard curve. The dynamic range is 31–2,000 pg/mL, with an analytical sensitivity of <12 pg/mL. Right panel: Kit recombinant human (Hu) tau protein standard compared to natural human tau serially diluted in Standard Diluent Buffer. The optical density of each dilution was plotted against the standard curve. Parallelism between the natural and recombinant protein indicates that the standard accurately reflects natural Hu tau content in samples.
Resources
Learning
Support
Related resources