The LifeScope™ Cluster is a multi-node, expandable, high-performance computing solution for the analysis and storage of data from you 5500 Series Genetic Analysis Systems. Pre-loaded with LifeScope™ Genomic Analysis Software, and performance-tuned for the analysis of 5500 Series Genetic Analysis Systems data, the LifeScope™ Cluster provides a flexible and scalable solution to meet the expanding computational needs of your organization.
Region Product Product Sku
United States LifeScope™ Cluster EL0100US 4466824
Europe LifeScope™ Cluster EL0100EU 4466825
Asia Pacific/Japan LifeScope™ Cluster EL0100AP 4466826
For additional product and pricing information:
have a rep contact me
Benefits of the LifeScope™ Cluster:
Enlarge Image
Figure 1: LifeScope™ Cluster (W=595mm × D=1,090mm × H=1,190mm) with pre-loaded and performance-tested LifeScope™ software.
  • High-performance computing, delivered—the LifeScope™ Cluster contains the latest computer components optimized for data-intensive next-generation sequence analysis
  • Integration is key—pre-loaded with LifeScope™ Genomic Analysis Software so you are ready to go as soon as you plug it in
  • Grows with your growing needs—expansion slots for additional compute nodes and long-term storage as your data analysis and managements needs expand
High-Performance Computing for 5500 Series SOLiD™ Data Analysis
The LifeScope™ Cluster (Figure 1) is a 4-node/48-core compute cluster with 2.66 GHz Intel Xeon X5650 processors, 24 Gigabytes (GB) of RAM per node, and an exceptionally fast internal network for optimized mapping and tertiary analysis of 5500 Series SOLiD™ data.  With approx. 9.5 Terabytes (TB) of usable storage, the entry-level model LifeScope™ Cluster is capable of processing large workload queues and running analyses in parallel.  Scyld ClusterWare cluster management software, with the new Insight interface, helps maintain compute efficiency.  The LifeScope™ Cluster is a convenient 24U (1.2m high) form factor, so it can be used in a laboratory setting as well as in a data center.
Integrated and Read to Go
Every LifeScope™ Cluster comes with LifeScope™ Genomic Analysis Software pre-installed and ready for licensing.  In addition, every cluster contains the necessary reference and other data to run the many genomic analyses enabled by the LifeScope™ software.  Finally, every cluster is tested prior to shipping to ensure maximum performance and reliability, so you can be confident that this high-performance solution will work out-of-the-box and as soon as you need it.
Scalable for Your Long-Term Needs
The LifeScope™ Cluster not only has the necessary compute specifications to run LifeScope™ Software and manage compute-intensive workloads, it also has the capacity to integrate with existing computing and storage infrastructures.  From the entry-level model, you can scale system performance by expanding up to 144 compute cores across 12 nodes, with up to 1.1 TB RAM per node.  For your growing storage and archiving needs, you can easily expand the cluster’s local storage up 108TB total memory.
A Community of Support
With more than 2,000 field personnel, Life Technologies has the most extensive network of dedicated Bioinformatics Scientists, Application Specialists, and Service Engineers to work with you, so that you can feel confident that we will be there when and where you need us most.
Learn More
For more information on the LifeScope™ Cluster, and to learn how to get your initial copy of LifeScope™ Genomic Analysis Software for free, please contact your local Life Technologies field Bioinformatics Scientist.