Search
One of the greatest challenges facing industrial R&D operations is the practical need to test a greater number of samples each day with no compromise on analytical quality.
Thermo Scientific ChemiSEM Technology goes a long way toward solving this challenge by fully integrating the EDS software needed to provide elemental analysis into the SEM microscope itself. The result is a single, seamless experience based on a simple and intuitive user interface that puts you in the best possible position to succeed.
You no longer need to learn and manage two separate software systems when conducting material analysis. ChemiSEM Technology quietly collects chemical information in the background, before you even need it, to deliver always-on live data that can help your laboratory process larger data volumes and create experimental datasets with higher statistical precision.
With ChemiSEM Technology, lab managers no longer need to spend valuable time and resources training their staff in specialized EDS systems. It is a versatile solution for higher-throughput analyses that provides the best of both worlds: a wide range of features that appeal to experienced users and simper configurations for those newer to the technique.
This is one way that ChemiSEM Technology unlocks valuable uptime on the microscope. Another is its unique, built-in reporting capabilities. Each session can be saved and exported for offline post processing anywhere.
After importing each data cube, you are presented with a wide range of options and features to help reanalyze data, adjust colors, extract spectra from smaller areas, or simply cross-check analyses with previously acquired data.
Experiment setup, image processing, and spectral analysis functions are intuitively grouped, making data management straightforward. And the system features a quadrant format and project tree to highlight the most important data.
Finally, a reporting function enables selected data to be easily saved and presented in a customized report.
With all these features and more, ChemiSEM Technology can help you make the most of your time.
For Research Use Only. Not for use in diagnostic procedures.