Find oligonucleotides built to your specifications here.
Our enhanced Standard Oligos ordering portal, now easier and faster.
DNA oligo products and services when you need them. Custom oligos are offered with standard deoxynucleotides, modified bases and 5' and 3'-modified nucleotides.
Sign In to Your Account | New Customer? Start here
Pricing | Ordering Details and Delivery Schedule | Contact Us | Oligo Quality Control | Benefits of web order
Service Line Up
Value Oligos
One price (pay per oligo)
with volume discounts
More about Value Oligos ›
Basic Entry
Upload Entry
  • 7-30 mers
  • 40 nmol scale
  • Desalted / No modifications
  • One-day delivery
Standard Oligos
Pay per base 
with 5', internal, or 3' modifications
Basic Entry
Upload Entry
  • 7-100 mers
  • 50 / 200 nmol , 1µ mol scale
  • Desalted / Cartridge / HPLC /PAGE
  • 5’ / 3’ / Internal modifications
Plate Oligos
One complete order with the same purification, synthesis scale, ship medium, and buffer
Upload Entry
  • More than 24 oligos / plate
  • 7-80 mer
  • 50 / 200nmol scale
  • Desalted / Cartridge
Delivery and Price Information
Product Information
RNA Oligos
Order RNA Oligos here
Resources