Our Newest Cellular Analysis Products and Technologies
On this page:
A new Mouse IFN-γ ELISA Kit for quantitative measurement of IFN-γ protein in mouse serum, plasma, or tissue culture supernatant is now available. The kit includes precoated plates and all the necessary reagents to analyze your samples.
- Learn More about all of our ELISA Kits
Protein disulfide isomerase (PDI) is a resident protein of the endoplasmic reticulum (ER) and can be used as an ER marker. New PDI antibodies are now available for detecting the ER and investigating misfolded protein pathways: PDI ABfinity™ Recombinant Rabbit Monoclonal Antibody, and PDI Recombinant Rabbit Oligoclonal Antibody, which is generated from a pool of clones. ABfinity recombinant antibodies are designed and manufactured for consistent performance, offering assurance that they will perform consistently in your application.
- Learn More About ABfinity Recombinant Rabbit Monoclonal Antibodies
Western blot analysis of whole cell extract using PDI Recombinant Rabbit Oligoclonal Antibody.
Molecular Probes flow cytometry antibodies in a trial size provide a convenient way for you to evaluate antibodies for inclusion in your flow cytometry panel. These fluorescent conjugates are excited at 405 nm to maximize use of the violet laser. The 25-test size is ideal for panel optimization and is available for Pacific Blue™ dye, Pacific Orange™ dye, and Qdot™ nanocrystal conjugates of our most popular antibodies.
Cellular labeling with the Qdot 655 conjugate of mouse anti–human CD4 antibody.
The popular pHrodo fluorogenic pH sensor for live cell studies is now available in a microscale size designed for three 20–100 μg labeling reactions. The labeling kit can stably attach pH-sensitive pHrodo Red dye to proteins, cells, antibodies, viruses, and almost any biomolecule, with excellent recovery. pHrodo Red avidin can be used to make conjugates of biotinylated molecules. The versatile pHrodo Red pH sensor dye can be detected with flow cytometry, microscopy, high-throughput screening, or high-content imaging platforms.
- Learn More About pHrodo Dyes
Visualizing virus internalization with pHrodo Red dye. CellLight™ MAP4-GFP, a BacMam virus, was labeled with the new pHrodo Red Microscale Labeling Kit. U2OS cells were incubated with labeled virus and imaged the next day. The pHrodo Red–labeled viruses retained functionality: green fluorescence along microtubules labeled with targeted GFP expressed from the BacMam construct can be seen, as can residual pHrodo Red staining from viral remnants.
Article Download
Get a copy of this article as it appears in the print version of BioProbes 66.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.