GIBCO Cell Culture Newsletter
GIBCO® Cell Culture Newsletter
Every other month we publish the GIBCO® Cell Culture Newsletter—your trusted resource for exciting developments and innovations in GIBCO® cell culture. With this newsletter, you can also take advantage of great deals and special offers on the cell culture products you use every day. To ensure that you continue to receive this vital information, click the Subscribe button below.
Share Your Protocol & Win!
Are you a loyal GIBCO® customer who has found better results by tweaking our protocol? Share your protocol with us and be entered in a drawing to become the GIBCO® Customer of the Month. The GIBCO® Customer of the Month will be randomly selected and will be announced in the next GIBCO® Cell Culture Newsletter.
So shine among your peers!  Send us your bio and a cool photo to be included in our next issue if you are the winner of the drawing. The GIBCO® Customer of the Month also wins a complimentary pass to the ISSCR 8th annual meeting OR a free Nintendo® Wii™ console.
Send us your protocol here!
Participation in the sweepstakes is open only to GIBCO® Cell Culture Newsletter subscribers. See more terms and conditions below.*  See terms and conditions below*
Issue 4: April 2010
NEWS AND PUBLICATIONS
Get GIBCO® products quickly and easily with Invitrogen's prepaid SimpleSci™ Card.
Sugii et al. demonstrate that human and mouse adipose-derived cells support feeder-independent induction
of pluripotent stem cells.
Aasen and Belmonte provide a protocol for the isolation and cultivation of human keratinocytes from
skin or plucked hair for the generation of induced pluripotent stem cells.
Chase et al. describe a novel serum-free medium for the expansion of human mesenchymal stem cells.
Introducing Invitrogen™ GIBCO® collagen-coated plates.
Watch the 2009 Cell Therapy Industry Summit videos.
Read about Invitrogen’s iPSC technologies in the latest issue of BioProbes.
New! ADME/TOX Services—See how we can support your drug development research.
FEATURED PRODUCTS
Hibernate®-A, Hibernate®-E—Preservation media for maintainingneuron viability in ambient CO2
StemPro® MSC SFM XenoFree---Xeno-free media for growth and expansion of human mesenchymal stem cells (MSCs)
One Shot FBS™—a simple concept with an amazing payoff
Getting ready to test?  Reserve your Fetal Bovine Serum today
iMATCH™ sera lot matching tool—The perfect match
KnockOut™ SR MEF Kit—Eliminate time spent sourcing MEFs
Tips and Tricks
  • CELLstart™ substrate should be diluted in DPBS that contains calcium and magnesium  (Cat. No. 14040141) because these inorganic salts help facilitate binding. CELLstart™ substrate can be used with human embryonic stem cells (hESCs), mesenchymal stem cells, neural stem cells, and  human induced pluripotent stem cells (iPSCs), and for the attachment of human feeder cells (e.g., foreskin fibroblasts).
  • Geltrex™ Basement Membrane Matrix can be used for more than just a coating layer for your hESCs! Geltrex™ matrix can also be used for cell invasion assays by applying a thick layer— 50–200 μL per cm2 --onto the growth surface. Incubate your plate for 30 minutes, and you're ready to go!
  • Cryopreservation of hESCs using KnockOut™ SR XenoFree. Prepare cryopreservation medium by supplementing KnockOut™ SR XenoFree Complete Medium with an additional 10% KnockOut™ SR XenoFree (to yield a final concentration of 25%) and 10% dimethyl sulfoxide (DMSO) cryoprotectant. Expect some cell death at recovery, and freeze hESCs at a higher density than would normally be passaged. (If cells are routinely passaged at a 1:5 dilution, a 1:3 or 1:4 dilution is recommended). .) Gently resuspend the cell pellet with cryopreservation medium without triturating. While transferring the contents into vials, invert the capped hESC tube regularlyroutinely to mix the cells. For best results, hESC vials should be Nalgene Cryo 1°C Freezing Containers).
Troubleshooting/FAQs
  1.    Can human MSCs be transferred directly into StemPro® MSC SFM XenoFree medium from FBS-supplemented medium?
Yes. Our studies show that MSCs transferred directly into StemPro® MSC SFM XenoFree demonstrate comparable expansion anddoubling times as DMEM + 10% MSC-qualified FBS.
  2.    What does it mean to be "XenoFree"?
XenoFree (XF) Component (Direct)—a component will be considered to be XenoFree (direct) if it has not been directly derived from animal tissue, cells, or body fluid except for tissues, cells, or body fluids of human origin. Components produced via bacterial, yeast, or other recombinant processes will also be considered to be XenoFree origin even if the manufacturing process includes animal components in the fermentation broth or process enzymes
  3.    What are the storage conditions for StemPro® MSC SFM and StemPro® MSC SFM XenoFree?
Kit Basal Medium Supplement Complete Medium
StemPro® MSC SFM 2-8°C until expiration date
  • 2-8°C not recommended
  • -5°C to -20°C
  • Recommend usage-size aliquots with minimal freeze-thaws
2-8°C, 4 weeks
StemPro® MSC SFM XenoFree 2-8°C until expiration date
  • 2-8°C not recommended
  • -5°C to -20°C
  • Recommend usage-size aliquots with minimal freeze-thaws
2-8°C, 4 weeks
Promotions
Request a free sample of Geltrex™ Reduced Growth Factor Matrix.
Get a free GIBCO® bottle weight.
Now through June 30, 2010, when you purchase $1,000 of qualifying GIBCO® FBS, receive your very own set of limited-edition Bendy Bottle Buddies!
Get an iPod from GIBCO®. Now through June 30, 2010, when you purchase $2,500 of GIBCO® FBS products in a single order, receive a free Apple® iPod nano® 8 GB device.
Events
Come see us at upcoming conferences and events:
ISSCR 8th Annual Meeting
June 16–19, 2010
Moscone West
San Francisco, CA USA
American Cytogenetic Conference
May 13–16, 2010
Niagara Falls, Ontario, Canada
Register now
Association of Genetic Technologists (AGT) 35th Annual
Meeting
June 3–5, 2010
Phoenix, AZ, USA
Register now
Latest Buzz
Need help finding the right cells?
The Cell Resource Center makes it easy.
Invitrogen™ iPhone™ apps and widgets
We offer an assortment of handy Invitrogen™ apps and widgets for your everyday
needs. From the convenient Everyday calculators app to the just-for-fun Play My DNAwidget, you’ll find many ways to quickly get the information you need.
*Terms and Conditions: Share Your Protocol & Win!
View complete terms and conditions.