Nitrocellulose membrane is a high quality membrane ideal for blotting of proteins and nucleic acids. The Nitrocellulose membrane is available in two pore sizes:
Catalog number
Pore Size
 0.2 µm     
Transfer of low molecular weight  proteins (<20 kDa) and  nucleic acids(<300 bp)
0.45 µm  
Transfer of most proteins (>20 kDa) and nucleic acids (>300 bp)
The major features of the Nitrocellulose membrane available from Invitrogen are:
  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Contains no support fabric or detergents
  • Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Provides high-sensitivity with low background
  • Supplied in a pre-cut, pre-assembled membrane/filter paper sandwich for convenience
Blotting Novex® Pre-Cast Gels
Western Blotting
Instructions are provided below for blotting Novex® Pre-Cast Gels using the XCell II™ Blot Module. For more information on the XCell II™ Blot Module, refer to the manual (IM-9051) available at or contact Technical Service.
If you are using any other blotting apparatus, follow the manufacturer’s recommendations.
Power Considerations for Blotting
During blotting, the distance traveled (gel thickness) between the electrodes is much lower than during electrophoresis requiring lower voltage and lower field strength (volts/distance). However, the cross sectional area of current flow is much greater requiring higher current.
Blotting power requirements depend on field strength (electrode size) and conductivity of transfer buffer. The higher the field strength and conductivity of the buffer, the higher is the current requirement (the current decreases during the run as the ions in the buffer polarize). It is important to use a power supply capable of accommodating the initial high current requirement.
Materials Supplied by the User
  • Blotting membranes
  • Filter paper (not needed if using Novex® pre-cut membrane/filter paper sandwiches)
  • XCell II™ Blot Module
  • Appropriate Transfer Buffer
  • Deionized water
Preparing Transfer Buffer For blotting Tris-Glycine, Tricine, and IEF Gels
We recommend using the Tris-Glycine Transfer Buffer. An alternate transfer protocol for IEF gels is provided.
If you are performing protein sequencing, alternate transfer buffers are listed.
Prepare 1000 ml of 1X Tris-Glycine Transfer Buffer using the Tris-Glycine Transfer Buffer (25X) as follows:
Tris-Glycine Transfer Buffer (25X)           40 ml
Methanol                                                   200 ml
Deionized Water                                       760 ml
Total Volume                                           1000 ml
See Recipe for a recipe of the Tris-Glycine Transfer Buffer, if you are preparing your own transfer buffer.
Preparing NuPAGE® Transfer Buffer
Prepare 1000 ml of 1X NuPAGE® Transfer Buffer using the NuPAGE® Transfer Buffer (20X) as follows:
  Reduced Samples Non-Reduced Samples
NuPAGE® Transfer Buffer (20X) 50 ml 50 ml
NuPAGE® Antioxidant 1 ml --
Methanol 100 ml* 100 ml*
Deionized Water 849 ml 850 ml
Total Volume 1000 ml 1000 ml
For blotting TBE and TBE-Urea Gels
Dilute the 5X TBE Running Buffer to 0.5X with deionized water.
Preparing Blotting Pads
Use about 700 ml of 1X Transfer Buffer to soak the pads until saturated. Remove the air bubbles by squeezing the pads while they are submerged in buffer. Removing the air bubbles is essential as they can block the transfer of biomolecules if they are not removed.
Preparing Transfer Membrane and Filter
Cut selected transfer membrane and filter paper to the dimensions of the gel or use Novex® pre-cut membrane/filter paper sandwiches.
  • Nitrocellulose—Place the membrane directly into a shallow dish containing 50 ml of 1X Transfer Buffer for several minutes.
  • Filter paper—Soak the filter paper briefly in 1X Transfer Buffer immediately prior to use.
  • Gel—Use the gel immediately following the run. Do not soak the gel in transfer buffer.
Western Transfer Using the XCell II™ Blot Module
Wear gloves while performing the blotting procedure to prevent contamination of gels and membranes, and exposure to irritants commonly used in electrotransfer.
Transferring One Gel
  1. After opening the gel cassette as described, remove wells with the Gel Knife.
  2. Place a piece of pre-soaked filter paper on top of the gel, and lay just above the slot in the bottom of the cassette, leaving the “foot” of the gel uncovered. Keep the filter paper saturated with the transfer buffer and remove all trapped air bubbles by gently rolling over the surface using a glass pipette as a roller.
  3. Turn the plate over so the gel and filter paper are facing downwards over a gloved hand or clean flat surface.
  4. Use the Gel Knife to push the foot out of the slot in the plate and the gel will fall off.
  5. When the gel is on a flat surface, cut the “foot” off the gel with the gel knife.
  6. Wet the surface of the gel with transfer buffer and position the pre-soaked transfer membrane on the gel, ensuring all air bubbles have been removed.
  7. Place another pre-soaked anode filter paper on top of the membrane. Remove any trapped air bubbles.
  8. Place two soaked blotting pads into the cathode (-) core of the blot module. The cathode core is the deeper of the two cores and the corresponding electrode plate is a darker shade of gray. Carefully pick up the gel membrane assembly and place on pad in the same sequence, such that the gel is closest to the cathode plate.
  9. Add enough pre-soaked blotting pads to rise to 0.5 cm over rim of cathode core. Place the anode (+) core on top of the pads. The gel/membrane assembly should be held securely between the two halves of the blot module ensuring complete contact of all components.
  10. Position the gel membrane sandwich and blotting pads in the cathode core of the XCell II™ Blot Module to fit horizontally across the bottom of the unit. There should be a gap of approximately 1 cm at the top of the electrodes when the pads and assembly are in place.
  11. Hold the blot module together firmly and slide it into the guide rails on the lower buffer chamber. The blot module will only fit into the unit one way, so the (+) sign can be seen in the upper left hand corner of the blot module. Properly placed, the inverted gold post on the right hand side of the blot module will fit into the hole next to the upright gold post on the right side of the lower buffer chamber.
  12. Place the gel tension wedge so that its vertical face is against the blot module. Lock the gel tension wedge by pulling the lever forward.
  13. Fill the blot module with 1X Transfer Buffer until the gel/membrane sandwich is covered in this buffer. Do not fill all the way to the top as this will only generate extra conductivity and heat.
  14. Fill the outer buffer chamber with deionized water by pouring approximately 650 ml in the gap between the front of the blot module and the front of the lower buffer chamber. The water level should reach approximately 2 cm from the top of the lower buffer chamber. This serves to dissipate heat produced during the run.
  15. Place the lid on top of the unit.
  16. With the power turned off, plug the red and black leads into the power supply. Refer to Recommended Transfer Conditions for transfer conditions.
Transferring Two Gels in One Blot Module
  1. Repeat Steps 1–6 above twice to make two gel-membrane assemblies.
  2. Place two pre-soaked pads on cathode shell of blot module. Place first gel-membrane assembly on pads in correct orientation, so gel is nearest cathode plate.Add another pre-soaked blotting pad on top of first membrane assembly.
  3. Position second gel-membrane assembly on top of blotting pad in the correct orientation so that the gel is nearest the cathode side.Proceed with steps 8–13 from Transferring One Gel.
  4. Refer to Recommended Transfer Conditions for transfer conditions.
Recommended Transfer Conditions
The transfer conditions for Novex® Pre-Cast Gels using the XCell II™ Blot Module are listed in the table below.
Note: The expected current listed in the table is for transferring one gel. If you are transferring two gels in the blot module, the expected current will double.
Transfer Buffer
Power Conditions
NuPAGE ® Novex Bis-Tris Gel
1X NuPAGE ® Transfer Buffer with 10% methanol*
0.1% NuPAGE ® Antioxidant for reduced samples
Nitrocellulose or PVDF
30 Volts constant for 1 hour
Expected Current
Start: 170 mA
End: 110 mA
NuPAGE ® Novex Tris-Acetate Gel
1X NuPAGE ® Transfer Buffer with 10% methanol*
0.1% NuPAGE ® Antioxidant for reduced samples
Nitrocellulose or PVDF
30 Volts constant for 1 hour
Expected Current
Start: 220 mA
End: 180 mA
Tris-Glycine Gel
Tricine Gel
1X Tris-Glycine Transfer Buffer with 20% methanol
Nitrocellulose or PVDF
25 V constant for 1-2 hours
Expected Current
Start: 100 mA
1X Tris-Glycine Transfer Buffer with 20% methanol
Nitrocellulose or PVDF
25 V constant for 1 hour
Expected Current
Start: 65-85 mA
0.7% Acetic acid pH 3.0
See for details on this alternate transfer protocol
Nitrocellulose or PVDF
10 V constant for 1 hour
Expected Current
Start: 65-85 mA
0.5X TBE Running Buffer
30 V constant for 1 hour
Expected Current
Start: 39 mA
End: 35 mA
TBE-Urea Gel
0.5X TBE Running Buffer
30 V constant for 1 hour
Expected Current
Start: 39 mA
End: 35 mA
DNA Retardation Gel
0.5X TBE Running Buffer
30 V constant for 1 hour
Expected Current
Start: 39 mA
End: 35 mA
Blotting IEF Gels
Novex® IEF Gels are composed of 5% polyacrylamide and are more susceptible to hydrolysis due to the heat generated with the recommended blotting protocol. The following protocol has been optimized to prevent hydrolysis and effective transfer of basic proteins due to the low pH of the transfer buffer.
  1. Chill 0.7% acetic acid which will be used later for transfer.
  2. After electrophoresis of the gel, equilibrate the gel in 0.7% acetic acid for 10 minutes. Tip:The 5% polyacrylamide gels are more sticky and difficult to handle than higher percentage polyacrylamide gels. To lift the gel from the equilibration solution, submerge the filter paper under the gel while the gel is floating in the equilibration solution. When the gel is in the correct position, lift up the filter paper to attach the gel to the filter paper. This prevents the gel from sticking to the filter paper before it is in the proper position and avoids handling of the gel.
  3. Assemble the gel/membrane sandwich as described previously, except in a reverse order so that the membrane is on the cathode (-) side of the gel. This is the opposite of a typical western blotting protocol, where the negatively charged protein will migrate toward the anode (+) during the transfer.
  4. Transfer for 1 hour at 10 V constant.
Blotting Native Gels
During SDS-PAGE all proteins have a net negative charge due to the SDS in the sample buffer and the running buffer. Proteins separated during native gel electrophoresis do not have a net charge which may cause problems during the transfer. Some native proteins may have a higher pI than the pH of the Tris-Glycine Transfer Buffer used in standard transfer protocols. Guidelines are provided below to increase the transfer efficiency of native proteins.
  • Increasing the pH of the transfer buffer to 9.2 (25 mM Tris Base, 25 mM glycine, pH 9.2), allows proteins with pI below 9.2 to transfer towards the anode electrode
  • Place a membrane on both sides of the gel if you are using the regular Tris-Glycine Transfer Buffer, pH 8.3. If there are any proteins that are more basic than the pH of the transfer buffer, they will be captured on the membrane placed on the cathode side of the gel
  • Incubate the gel in 0.1% SDS for 15 minutes before blotting with Tris-Glycine Transfer Buffer. The small amount of SDS will render enough charge to the proteins so they can move unidirectionally towards the anode and in most cases will not denature the protein
Native proteins may diffuse out of the membrane into the solution during the blocking or antibody incubation steps, as the native proteins tend to be more soluble. To prevent diffusion of the proteins out of the membrane, we recommend fixing the proteins to the membrane by air drying the membrane or incubating the membrane in 5-10% acetic acid for 15 minutes followed by rinsing the membrane with deionized water and then air drying.
By performing any of these two fixing methods the proteins will be sufficiently unfolded to expose hydrophobic sites and bind more efficiently to the membrane.