Search
| Organism Type | Rat | The rat pituitary cell line GH4C1 secretes granins (chromogranin B and secretogranin II) and prolactin by the regulated secretory pathway. The intracellular storage of prolactin is preferentially induced by hormone treatment with estradiol, insulin, and epidermal growth factor. |
| Tissues | Pituitary | |
| Phenotype | Adherent | |
| Primary | yes | |
| Application | Stable Cell Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||