3a. COL116546-GTS-Accufill-New-banner-hero-graphic-needed-for-webpage-1600x422-F
Important
Before upgrading software or firmware, review the release notes and user guide for recalibration requirements. Changes may impact analytically validated workflows. Customers operating under analytically validated workflows must follow current Instructions for Use (IFU) and/or perform analytical validation according to their standard operating procedures.
QuantStudio 12K Flex Real-Time PCR Software
The instrument operating software enables instrument operation, run setup, and basic data analysis. The software can be run with security, audit, and e-signature (SAE) enabled.
The companion OpenArray Accufill software is also available to assist with OpenArray plate preparation.
Get software
Diomni Design and Analysis Software 3.x (DA3)
Diomni Design and Analysis 3.x (DA3) is designed to complement the instrument software by extending the following advanced data analysis and results review capabilities across the QuantStudio qPCR instrument family, including QuantStudio 12K systems:
Key capabilities
  • CNV and genotyping analysis
  • Multi-plate data analysis
  • Improved Ct algorithm performance
  • Advanced spectral conditioning (cross-talk reduction)
Access options
  • Desktop (Win & Mac)
  • On-Premises (Server + browser access)
  • Cloud (hosted on ThermoFisher Connect)
Desktop  On-premises  Cloud
Note: DA3 does not support instrument control or SAE (security, audit, e-signature) for QuantStudio 12K Flex System.
How to access software
  1. For Cloud: Select to launch the application directly.
  2. For Desktop or On-Premises: Select your install option (Desktop or On-Premises).
  3. Sign in with your thermofisher.com account (create one if needed).
  4. Access the Thermo Fisher Connect folder and download your software.
For Research Use Only. Not for use in diagnostic procedures.