Search
Join the Conversation
Thermo Scientific Helios 5 PFIB Laser Systems combine plasma focused ion beam milling with femtosecond laser ablation and SEM (scanning electron microscopy) imaging. This “TriBeam” combination enables high-resolution imaging and analysis with in situ ablation capability, offering unprecedented material removal rates for fast millimeter-scale characterization at nanometer resolution.
The femtosecond laser can cut many materials at rates that are orders of magnitude faster than a typical FIB. A large cross-section (hundreds of micrometers) can be created in less than five minutes. Because the laser has a different removal mechanism (ablation versus the ion sputtering of FIB), it can easily process challenging materials, such as non-conductive or ion-beam-sensitive samples.
The extremely short duration of the femtosecond laser pulses introduces almost no artifacts such as heat impact, microcracking, melting, or those typical of traditional mechanical polishing. In most cases, the laser-milled surfaces are clean enough for direct SEM imaging and even for surface-sensitive techniques such as electron backscatter diffraction (EBSD) mapping.
We offer a broad product portfolio and advanced automation capabilities for applications such as transmission electron microscopy (TEM) sample preparation, atom probe tomography (APT) sample preparation, and 3D structural analysis.
Built on the proven Helios 5 DualBeam platform, these instruments incorporate a suite of state-of-the-art technologies to provide high-performance, high-resolution transmission electron microscopy (TEM) and atom probe tomography (APT) sample preparation and extremely high-resolution SEM imaging with precise materials contrast.
When combined with Thermo Scientific Auto Slice & View Software, TriBeam instruments provide 3D insight into sample structure by selectively removing (milling) the material for subsurface characterization. Digital reconstruction generates multi-modal 3D datasets that can consist of a variety of signals, including backscattered electron (BSE) imaging for maximum materials contrast, energy dispersive spectroscopy (EDS) for compositional information, and electron backscatter diffraction (EBSD) for microstructural and crystallographic information. The SEM capability of TriBeam instruments offers nanoscale details across a wide range of working conditions, from structural information obtained at 30 keV in STEM mode to charge-free, detailed surface information at lower energies. With unique in-lens detectors, TriBeam systems are designed for simultaneous acquisition of angular/energy-selective secondary-electron and BSE data. Fast, accurate, and reproducible results are provided by our unique SEM column design, which features fully automated lens alignments.
Thermo Fisher Scientific offers a broad range of (P)FIB-SEM systems with fully integrated femtosecond lasers. This TriBeam product family consists of three models, all featuring the best-in-class Thermo Scientific Elstar SEM Column, a high-precision 150 mm piezo stage, and a large chamber. The main difference between these models is the FIB column:
Millimeter-scale cross sections with up to 15,000x faster material removal than a typical focused ion beam.
Data acquisition for much larger volumes within a shorter amount of time.
Same coincident point for all tree beams (SEM / (P)FIB / laser) enables accurate and repeatable cut placement and 3D characterization.
Extraction of subsurface TEM lamella or chunks for 3D analysis.
Includes non-conductive or ion-beam-sensitive samples.
No need to transfer samples between different instruments for cross-sectioning and imaging.
High-quality TEM and APT sample preparation and high-resolution imaging capabilities.
| Laser specifications | ||
|---|---|---|
| Laser integration |
| |
| First Harmonic |
| 1030 nm (IR) <280 fs |
| Second Harmonic |
| 515 nm (green) |
| Optics |
|
|
| Repetition rate | • 1 kHz – 1 MHz | |
| Beam position accuracy | • <250 nm | |
| Protective shutter | • Automated SEM/PFIB protective shutter | |
| Software | • Laser control software | |
| Safety | • Interlocked laser enclosure (Class 1 laser safety) | |
Register for our recorded webinar and learn how the combination of fs-Laser and PFIB provides mm-scale subsurface and 3D analysis at nm resolution and enables new workflows such as fast characterization of air sensitive samples and multi-scale correlative microscopy with deep subsurface sample extraction.
Register for our live webinar and learn how leading research labs are using our new Thermo Scientific Helios 5 Laser PFIB and Thermo Scientific Helios 5 Hydra DualBeam to advance their materials characterization.
Register for our recorded webinar and learn how the combination of fs-Laser and PFIB provides mm-scale subsurface and 3D analysis at nm resolution and enables new workflows such as fast characterization of air sensitive samples and multi-scale correlative microscopy with deep subsurface sample extraction.
Register for our live webinar and learn how leading research labs are using our new Thermo Scientific Helios 5 Laser PFIB and Thermo Scientific Helios 5 Hydra DualBeam to advance their materials characterization.
El control y garantía de calidad son esenciales en la industria moderna. Ofrecemos una gama de herramientas de EM y espectroscopía para el análisis multiescala y multimodal de defectos, lo que le permite tomar decisiones fiables e informadas para el control y la mejora de procesos.
Se investigan nuevos materiales a escalas cada vez más pequeñas para lograr el máximo control de sus propiedades físicas y químicas. La microscopía electrónica proporciona a los investigadores información clave sobre una amplia variedad de características materiales a escala nanométrica.
Caracterización de materiales en 3D
El desarrollo de materiales suele requerir caracterización en 3D en varias escalas. Los instrumentos DualBeam permiten el corte en secciones en serie de grandes volúmenes y la posterior adquisición de imágenes SEM a escala de nanómetro, las cuales se pueden procesar en reconstrucciones 3D de la muestra de alta calidad.
Preparación de muestras (S)TEM
Los microscopios DualBeam permiten la preparación de muestras ultrafinas de alta calidad para el análisis (S)TEM. Gracias a la automatización avanzada, los usuarios con cualquier nivel de experiencia pueden obtener resultados de nivel experto para una amplia gama de materiales.
Preparación de muestras de APT
La tomografía de sonda atómica (APT) proporciona un análisis de composición de materiales en 3D con resolución atómica. La microscopía Focused ion beam (FIB) es una técnica esencial para la preparación de muestras de alta calidad, orientación y sitio específico para la caracterización de APT.
Corte transversal
El corte transversal proporciona una visión adicional, ya que descubre información de la subsuperficie. Los instrumentos DualBeam tienen columnas FIB para poder realizar el corte transversal con alta calidad. Con la automatización, se puede realizar el procesamiento de muestras de alto rendimiento sin supervisión.
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.
Caracterización de materiales en 3D
El desarrollo de materiales suele requerir caracterización en 3D en varias escalas. Los instrumentos DualBeam permiten el corte en secciones en serie de grandes volúmenes y la posterior adquisición de imágenes SEM a escala de nanómetro, las cuales se pueden procesar en reconstrucciones 3D de la muestra de alta calidad.
Preparación de muestras (S)TEM
Los microscopios DualBeam permiten la preparación de muestras ultrafinas de alta calidad para el análisis (S)TEM. Gracias a la automatización avanzada, los usuarios con cualquier nivel de experiencia pueden obtener resultados de nivel experto para una amplia gama de materiales.
Preparación de muestras de APT
La tomografía de sonda atómica (APT) proporciona un análisis de composición de materiales en 3D con resolución atómica. La microscopía Focused ion beam (FIB) es una técnica esencial para la preparación de muestras de alta calidad, orientación y sitio específico para la caracterización de APT.
Corte transversal
El corte transversal proporciona una visión adicional, ya que descubre información de la subsuperficie. Los instrumentos DualBeam tienen columnas FIB para poder realizar el corte transversal con alta calidad. Con la automatización, se puede realizar el procesamiento de muestras de alto rendimiento sin supervisión.
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.