Search
The discovery of novel single nucleotide polymorphisms (SNPs) is critical for plant genotyping in that it helps to provide candidate markers for marker-assisted selection programs in key crops. Genomic approaches are useful for accelerating variety development and crop improvement. Genomic tools can assist in the detection of QTLs, favorable and deleterious traits and identification of favorable alleles. Thermo Fisher Scientific offers a range of genomic tools and resources to help facilitate your plant breeding and production goals.
Our array-based genotyping solutions are ideal for the identification, validation and screening of complex genetic traits in plants and animals. Choose from our wide variety of predesigned arrays or customize an array specifically for your study.
Product | No. of markers | Array Format | Cat. No. | Price |
| Axiom Citrus HD Genotyping Array (Citrus15 AX1) | 702,244 | 96-array | 550729 | Contact a sales representative › |
| Axiom Citrus HD Genotyping Array (Citrus15 AX2) | 702,198 | 96-array | 550730 | Contact a sales representative › |
| Axiom Citrus Genotyping Array | 58,433 | 384-array | 550775 | Contact a sales representative › |
| Axiom Peanut Genotyping Array | 47,837 | 384-array | 550846 | Contact a sales representative › |
| Axiom Peanut Genotyping Array | 47,837 | Mini 96-array | 550865 | Contact a sales representative › |
| Axiom Rose Genotyping Array | 137,786 | 96-array | 550450 | Contact a sales representative › |
| Axiom Rye Genotyping Array | 600,843 | 96-array | 550535 | Contact a sales representative › |
| Axiom Strawberry Genotyping Array (i35) | 34,260 | 384-array | 550767 | Contact a sales representative › |
| Axiom Strawberry Genotyping Array (i35) | 34,260 | Mini 96-array | 550827 | Contact a sales representative › |
| Axiom Tomato Genotyping Array | 51,214 | 96-array | 550497 | Contact a sales representative › |
These assays provide a highly flexible technology for detection of polymorphisms within any genome. Create your own assays by submitting target sequences to our secure assay design pipeline using the Applied Biosystems Custom TaqMan Assay Design Tool. This pipeline has successfully generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. TaqMan Assays can be run on any of our real-time PCR instruments or used as an endpoint assay on other third-party systems, including the Array Tape platform.