Search
Silencer Select siRNAs to 10 different targets (BUB1B, AURKB, WEE1, PLK1, FDFT1, HMGCR, LDLR, AURKA,CYP51A1, SQLE) and siRNAs from two other suppliers to the same 10 different targets were individually transfected into HeLa cells in triplicate at the indicated siRNA concentration. mRNA knockdown levels were tested 48 h later. Average percent mRNA remaining is shown for each set of siRNAs.