Bull sperm. MitoTracker® Green FM and Hoechst 33342.
Live bull sperm stained simultaneously with MitoTracker® Green FM (Cat. no. M7514) and Hoechst 33342 (Cat. no. H1399, H3570, H21492), as reported in (Mol Reprod Dev 47, 79 (1997)) . The image contributed by Peter Sutovsky, Oregon Regional Primate Resource Center, Oregon Health Sciences University, and used with the permission of Wiley-Liss, Inc., a subsidiary of John Wiley & Sons, Inc.
Live bull sperm stained simultaneously with MitoTracker® Green FM (Cat. no. M7514) and Hoechst 33342 (Cat. no. H1399, H3570, H21492), as reported in  (Mol Reprod Dev 47, 79 (1997)) . The image contributed by Peter Sutovsky, Oregon Regional Primate Resource Center, Oregon Health Sciences University, and used with the permission of Wiley-Liss, Inc., a subsidiary of John Wiley & Sons, Inc.
Related Products
Related Images