Search
High-quality protein is modified with Sulfo-SMCC crosslinker to produce the maleimide-activated carrier protein, which is supplied lyophilized. Addition of peptides or other molecules containing reduced sulfhydryl groups results in efficient and specific reaction to form stable thioether bonds. Each carrier protein molecule contains several hundred maleimide activations, allowing conjugation of many peptide hapten molecules per immunogenic carrier protein molecule.