Search
ELISA was performed to detect the antigen MIP-beta by serial dilution (3 fold starting at 1 ug/ml) of Goat anti-Mouse IgG (H+L) Superclonal? Secondary Antibody, Alkaline Phosphatase conjugate (A28184). A non-linear regression analysis was performed and EC50 determined to be 0.045 ug/ml.