Alexa Fluor 532 dye
Alexa Fluor 532 is a yellow-fluorescent dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Alexa Fluor 532 dye is pH-insensitive over a wide molar range, helping ensure that you get stable signals in live-cell imaging applications.
Alexa Fluor 532 conjugates exhibit an excitation maximum at 532 nm and emission maximum at 554 nm. It is particularly useful in multicolor applications, including super-resolution microscopy using dSTORM.
We offer Alexa Fluor 532 dye conjugated to a variety of antibodies and peptides optimized for cellular labeling and detection. In addition, reactive dye forms and protein labeling kits are available to enable you to generate your own antibody conjugates or probes.
Alexa Fluor 532 dye dashboard
Initial brightness |
Alexa Fluor 532 dye is used in stochastic optical reconstruction microscopy (STORM)—as an activator in nSTORM and a reporter in dSTORM.
|
|
Photostability in buffer |
||
Photostability in antifade |
||
Performance in SRM |
| 532 | TRITC | 532 | 554 | ||
Laser line |
Common filter set |
Excitation max |
Alexa Fluor 532 dye products
We offer a broad range of Alexa Fluor 532 products including labeling kits, secondary antibodies, and bioconjugates:
Human mononuclear cells were stained with mouse anti human CD4 biotin, washed, stained with streptavidin, Alexa Fluor 532 conjugate, washed, and analyzed by flow cytometry using 532 nm excitation.
Alexa Fluor 532 reactive dyes
Select dye-labeling chemistries for conjugation with the following reactive groups:
Alexa Fluor 532 protein/antibody labeling kits
Select an optimized labeling kit for your target antibody or protein:
| Target protein | Kit | Cat. No. |
|---|---|---|
| Site-specific antibody labeling | ||
| <1–20 µg mouse IgG1 | Zenon Alexa Fluor 532 Mouse IgG1 Labeling Kit | Z25003 |
| <1–20 µg rabbit IgG | Zenon Alexa Fluor 532 Rabbit IgG Labeling Kit | Z25303 |
| Conventional antibody/protein labeling | ||
| 100 µg antibody/protein | Alexa Fluor 532 Antibody/Protein Labeling Kit | A20182 |
| 1 mg antibody/protein | Alexa Fluor 532 Antibody/Protein Labeling Kit | A10236 |
Alexa Fluor dye family
Illuminating research for over 15 years
Alexa Fluor dyes are among the most trusted fluorescent dyes available today. In addition to superior performance, you’ll get a technical support team experienced in problem-solving, >50,000 published references, application and experimental tips, and protocols to help with experimental planning. For an outstanding research experience, choose Alexa Fluor dyes.
- Learn more about the brightness of Alexa Fluor dyes
- Learn more about the photostability of Alexa Fluor dyes
- Read all about Alexa Fluor dyes in the Molecular Probes Handbook
For Research Use Only. Not for use in diagnostic procedures.