Search
H9 human embryonic stem cells (ESCs) were cultured on mouse embryonic fibroblasts (MEFs) with 20% ESC-Qualified Fetal Bovine Serum (FBS) or 20% KnockOut SR – Multi-Species. The mean viable cell numbers were recorded to an extrapolated growth curve for ESC-Qualified FBS-based media and KSR – Multi-Species-based media. Proliferation of human ESCs was significantly higher in KSR – Multi-Species over 3 passages.