Search
The IL-12 p40 Human Direct ELISA Kit is designed to quantify protein levels of human IL-12 p35/p40 heterodimer, also known as IL-12p70, and also detects the IL-12 p40/p40 homodimer, also known as IL-12p80.
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›