The 2720 Thermal Cycler will be discontinued December 31, 2018.
The product replacing the 2720 Thermal Cycler is the MiniAmp Thermal Cycler.
Applied Biosystems 2720 Thermal Cycler
질병 진단을 위해 특정 유전자(DNA)의 증폭에 사용하는 자동장치로서, 내부 Peltier heating/ cooling 장비를 포함하는96-well 샘플 블록을 가집니다.
샘플을 0.2mL의 reaction tube나 96-well reaction plate에 loading 하여 유전자를 증폭시킵니다.
(유전자증폭장치, 수허 12-259호, GeneAmp 2720 Thermal Cycler)
"본 제품은 의료기기이며, 사용시 주의사항사용방법을 잘 읽고 사용하십시오."
Industry-standard PCR technology
Our newly launched MiniAmp Thermal Cycler, like its 2720 Thermal Cycler predecessor, continues the Applied Biosystems tradition of reliability and performance that customers around the world have come to expect from Applied Biosystems thermal cyclers.
Spacing saving footprints
The 2720 Thermal Cycler fits almost anywhere with dimensions of only 21 cm (8.3 in.) wide and 36 cm (14.2 in.) deep. And because its cooling vents are in the back, you can safely squeeze it into tight spaces to save your valuable bench space.
The footprint for the MiniAmp Thermal Cycler is even smaller (7.5 in (19 cm) wide), and at an entry-level price, they are perfect for every lab bench.
* GeneAmp 2720 Thermal Cycler 사용설명서.pdf
GeneAmp 2720 Thermal Cycler 사용설명서
정격사양 100∼240V, 50/60Hz, 420VA
치수 및 중량 Height: 22cm, Width: 21cm, Depth: 36cm, Weight: 6.1kg
Well 수 96-well
온도정확도 ±0.5℃
온도균일성 ±0.5℃
온도범위 4.0℃∼99.9℃
가열/냉각 온도 ramp rate Heating rate: >1.25 ℃/Sec Cooling rate: >1.55 ℃/Sec