The Ion GeneStudio S5 Series is flexibly designed to enable a broad range of targeted NGS applications with industry-leading speed and scalability. Select from five different sequencing chips to sequence a throughput range from 2M to 130M reads per run. Simply choose the chip size and the instrument that matches your throughput and application needs.
Get a quote    Product bulletin    View specifications
Meet the Ion GeneStudio S5 system
Which Ion GeneStudio S5 System is right for you?
Ion GeneStudio S5 System
Ion S5 and S5 XL NGS Sequencing Systems
Inquire
Get started with targeted NGS
Ion GeneStudio S5 Plus System
Ion S5 and S5 XL NGS Sequencing Systems
Discover
Choose flexibility to support small and large projects
Ion GeneStudio S5 Prime System
Decide
Save time with greater throughput
Ion chip compatibility
Ion 510 Chip
Ion 520 Chip
Ion 530 Chip
Ion 540 Chip
Ion 510 Chip
Ion 520 Chip
Ion 530 Chip
Ion 540 Chip
Ion 550 Chip
Ion 510 Chip
Ion 520 Chip
Ion 530 Chip
Ion 540 Chip
Ion 550 Chip
Maximum throughput/day (chip type)
15 Gb (1 Ion 540 Chip)
30 Gb (2 Ion 540 chips)
50 Gb (2 Ion 550 chips)
Speed
19 hrs (Ion 540 Chip)
10 hrs (Ion 540 Chip)
11.5 hrs (Ion 550 Chip)
6.5 hrs (Ion 540 Chip)
8.5 hrs (Ion 550 Chip)
Learn about the simple workflow of the Ion GeneStudio S5 Series
Ion S5 sequencer 와 Ion S5 XL sequencer 장비는 샘플 준비작업을 거친 DNA template 을 이용하여, DNA 염기서열검사반응에서 각각의 뉴클레오타이드 결합 중 발생되는 수소이온을 반도체칩(semi‐conducting chip)으로 측정해 염기서열을 분석하는 염기서열검사기입니다. 본 제품은 사람의 유전자 중 특정 여러 위치에서의 유전자 시퀀스분석에 사용합니다.
(유전자서열검사기, 수인 15-4212 호, Ion S5 Sequencer / Ion S5 XL Sequencer)
"본 제품은 의료기기이며, 사용방법과 사용시 주의사항을 잘 읽고 사용하십시오."
# 명칭 제품번호 설명 특성
1
Ion Server
(S5 XL 구성품)
A28563
sequencing 이 완료된 후 data 분석을 위한 외부컴퓨터.
data 분석을 통한 1 차 결과분석을 수행한다.
-100-240VAC, 50/60Hz, 6.7A
-DELL PowerEdge T630 tower
-Intel Xeon E5-2680 v3 processors
-Memory : 128GB
-치수:
56.6cm(H)x56.6cm(L)x21.6cm(W)
※ 동일사양 이상에 한하여 호환가능
2 Ion Chef Instrument 4484178
유전자 샘플 준비단계에서 사용되는 자동샘플준비장비.
library DNA 를 ISP 에 부착시켜 증 폭 후 농축 ISP 상태로 enrich 거쳐 sequencing 준비를 한다.
-100-240VAC, 50/60Hz, 12A
-치수:
56.1cm(H)x 70.0cm(D)x 71.4cm(W)
-중량: 68.2kg
3 Ion 520 Chip Kit A27762
준비한 샘플을 주입하여 분석하는 반도체칩.
1 개 chip 내 약 1 천만개의 well 을 보유.
-보관온도: 15~30℃
-약 1 천만개 well / 1 chip
- 8 chips / 1 kit
4 Ion 530 Chip Kit A27763
준비한 샘플을 주입하여 분석하는 반도체칩.
1 개 chip 내 약 4 천만개의 well 을 보유.
-보관온도: 15~30℃
-약 4 천만개 well / 1 chip
- 8 chips / 1 kit
5 Ion 540 Chip Kit A27766
준비한 샘플을 주입하여 분석하는 반도체칩.
1 개 chip 내 약 1 억 6 천만개의 well 을 보유.
-보관온도: 15~30℃
-약 1 억 6 천만개 well / 1 chip
- 8 chips / 1 kit
Speed and flexibility improve lab competitiveness
“In my institute, many customers outsource for exome and whole genome studies.  They come to me for speed, and flexibility for different gene panels, tailored to disease areas of interest.  This is the most elegant workflow on the market today, and I can now offer a range of applications and deliver the data in the fastest turnaround time in the market, at costs that are competitive.” 
—Morten Dunoe, Lab Manager, Molecular Genetic Laboratory, Department of Clinical Genetics, Rigshospitalet, Copenhagen University Hospital, Denmark
See what you can do—delve into application notes
Cancer Genomics Transcriptomics Research
Cancer genomics and transcriptomics research
Low Frequency Mutations Cell Free DNA
Low-frequency mutations from cell-free DNA
Reproductive Genomics Inherited Disease Research
Reproductive genomics and inherited disease research
Rapid Sequencing Microorganisms
Rapid sequencing of microorganisms
Resources