Search
A-C: After floor plate specification of H9 ESCs, cells express floor plate marker FOXA2 and rostral marker OTX2. D-E: The specified cells are positive for midbrain marker LMX1A (>80% by Cellomics analysis) but negative for neural stem cell marker SOX1.