Human Hematopoietic Lineage Antibody (22777872) in Flow
Normal human peripheral blood cells were stained with Anti-Human CD34 Alexa Fluor® 647, Anti-Human CD59 (Protectin) PE (cat. 12-0597), and the Human Hematopoietic Lineage FITC Cocktail. Total viable cells were used for analysis. Cells in the right dot plot are gated on lineage negative events, depicted in the left histogram.
Normal human peripheral blood cells were stained with Anti-Human CD34 Alexa Fluor® 647, Anti-Human CD59 (Protectin) PE (cat. 12-0597), and the Human Hematopoietic Lineage FITC Cocktail. Total viable cells were used for analysis. Cells in the right dot plot are gated on lineage negative events, depicted in the left histogram.
Related Products
Related Images