LentiArray Cas9 Lentivirus map
A CRISPR-Cas9 targeted double-strand break. Cleavage occurs on both strands, 3 bp upstream of the NGG in the protospacer adjacent motif (PAM) sequence on the 3´ end of the target sequence.
A CRISPR-Cas9 targeted double-strand break. Cleavage occurs on both strands, 3 bp upstream of the NGG in the protospacer adjacent motif (PAM) sequence on the 3´ end of the target sequence.
Related Products
Related Images