CD69 Antibody (62-0691-82) in Flow
BALB/c mouse splenocytes were unstimulated (left) or stimulated overnight with Con A (Product ## 00-4978) (right). Cells were then stained with CD25 Monoclonal Antibody, PE (Product ## 12-0251) and 0.25 µg of CD69 Monoclonal Antibody, Super Bright 436. Cells in the lymphocyte gate were used for analysis.
BALB/c mouse splenocytes were unstimulated (left) or stimulated overnight with Con A (Product ## 00-4978) (right). Cells were then stained with CD25 Monoclonal Antibody, PE (Product ## 12-0251) and 0.25 µg of CD69 Monoclonal Antibody, Super Bright 436. Cells in the lymphocyte gate were used for analysis.
Related Products
Related Images