Indirect ELISA analysis of Angptl4 ABfinity™ Recombinant Rabbit Monoclonal Antibody – Purified
Indirect ELISA was performed to detect Angptl4 conjugated peptide coated onto the plate. Various dilutions of Angptl4 ABfinity™ Recombinant Rabbit Monoclonal Antibody (Cat. No.701155) was used followed by secondary antibody Goat anti Rabbit HRP. A non-linear regression analysis was performed (4 PL) and LOD and LOQ for the antibody was determined.
Indirect ELISA was performed to detect Angptl4  conjugated peptide coated onto the plate. Various dilutions of Angptl4 ABfinity™ Recombinant Rabbit Monoclonal Antibody (Cat. No.701155) was used followed by secondary antibody Goat anti Rabbit HRP. A non-linear regression analysis was performed (4 PL) and LOD and LOQ for the antibody was determined.
Related Products
Related Images